Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193105405-193105674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX2
Entrez Gene ID
51022
Gene Name
glutaredoxin 2
Gene Aliases
CGI-133, GRX2
UniGene
Hs.458283
Gene Chromosome Location
Chr.1: 193096465-193106535 on Build GRCh38
Transcripts
NM_001243399.1 NM_001319291.1 NM_016066.4 NM_197962.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750712 c.118A>T p.R40W Chr.1: 193105521-193105521 on Build GRCh38
2124131 c.83G>A p.R28H Chr.1: 193105556-193105556 on Build GRCh38
6459841 c.83G>C p.R28P Chr.1: 193105556-193105556 on Build GRCh38
424876 c.117G>A p.A39A Chr.1: 193105522-193105522 on Build GRCh38
7129689 c.22G>A p.V8M Chr.1: 193105617-193105617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040484958 Chr.1: 193105458-193105458 on Build GRCh38
rs984751423 Chr.1: 193105622-193105622 on Build GRCh38
rs994128890 Chr.1: 193105433-193105433 on Build GRCh38
rs1021340973 Chr.1: 193105549-193105549 on Build GRCh38
rs759448025 Chr.1: 193105534-193105534 on Build GRCh38
rs750044838 Chr.1: 193105581-193105581 on Build GRCh38
rs34533242 Chr.1: 193105504-193105504 on Build GRCh38
rs774481041 Chr.1: 193105522-193105522 on Build GRCh38
rs769137129 Chr.1: 193105589-193105589 on Build GRCh38
rs747307173 Chr.1: 193105534-193105534 on Build GRCh38
rs775290178 Chr.1: 193105647-193105647 on Build GRCh38
rs746183694 Chr.1: 193105513-193105513 on Build GRCh38
rs761942184 Chr.1: 193105526-193105526 on Build GRCh38
rs1075660 Chr.1: 193105562-193105562 on Build GRCh38
rs202205053 Chr.1: 193105556-193105556 on Build GRCh38
rs924140482 Chr.1: 193105434-193105434 on Build GRCh38
rs966255415 Chr.1: 193105574-193105574 on Build GRCh38
rs554713403 Chr.1: 193105451-193105451 on Build GRCh38
rs757897856 Chr.1: 193105507-193105507 on Build GRCh38
rs770228849 Chr.1: 193105473-193105473 on Build GRCh38
rs752638722 Chr.1: 193105553-193105553 on Build GRCh38
rs768748717 Chr.1: 193105483-193105483 on Build GRCh38
rs1004793672 Chr.1: 193105486-193105486 on Build GRCh38
rs1075661 Chr.1: 193105599-193105599 on Build GRCh38
rs772251614 Chr.1: 193105615-193105615 on Build GRCh38
rs958583412 Chr.1: 193105532-193105532 on Build GRCh38
rs756927784 Chr.1: 193105576-193105576 on Build GRCh38
rs10921310 Chr.1: 193105521-193105521 on Build GRCh38
rs756838186 Chr.1: 193105512-193105512 on Build GRCh38
rs751239359 Chr.1: 193105511-193105511 on Build GRCh38
rs773766463 Chr.1: 193105649-193105649 on Build GRCh38
rs200163239 Chr.1: 193105427-193105427 on Build GRCh38
rs373761376 Chr.1: 193105502-193105502 on Build GRCh38
rs545839664 Chr.1: 193105516-193105516 on Build GRCh38
rs978254313 Chr.1: 193105428-193105428 on Build GRCh38
rs371731758 Chr.1: 193105491-193105491 on Build GRCh38
rs769547449 Chr.1: 193105635-193105635 on Build GRCh38
rs746379679 Chr.1: 193105431-193105431 on Build GRCh38
rs772360130 Chr.1: 193105527-193105527 on Build GRCh38
rs770314886 Chr.1: 193105515-193105515 on Build GRCh38
rs921903949 Chr.1: 193105461-193105461 on Build GRCh38
rs748187752 Chr.1: 193105606-193105606 on Build GRCh38
rs780781619 Chr.1: 193105580-193105580 on Build GRCh38
rs756519732 Chr.1: 193105583-193105583 on Build GRCh38
rs887693150 Chr.1: 193105476-193105476 on Build GRCh38
rs141243509 Chr.1: 193105544-193105544 on Build GRCh38
rs1048114120 Chr.1: 193105456-193105456 on Build GRCh38
rs527610252 Chr.1: 193105616-193105616 on Build GRCh38
rs764045332 Chr.1: 193105648-193105648 on Build GRCh38
rs146314703 Chr.1: 193105587-193105587 on Build GRCh38
rs574807010 Chr.1: 193105501-193105501 on Build GRCh38
rs766340346 Chr.1: 193105498-193105498 on Build GRCh38
rs760094514 Chr.1: 193105481-193105481 on Build GRCh38
rs753616661 Chr.1: 193105499-193105499 on Build GRCh38
rs924087981 Chr.1: 193105566-193105566 on Build GRCh38
rs374740662 Chr.1: 193105508-193105508 on Build GRCh38
rs139755695 Chr.1: 193105594-193105594 on Build GRCh38
rs1002589853 Chr.1: 193105519-193105519 on Build GRCh38
rs751023523 Chr.1: 193105568-193105568 on Build GRCh38
rs780407209 Chr.1: 193105469-193105470 on Build GRCh38
rs944774720 Chr.1: 193105448-193105448 on Build GRCh38
rs749824478 Chr.1: 193105588-193105588 on Build GRCh38
rs368036406 Chr.1: 193105530-193105530 on Build GRCh38
rs762901131 Chr.1: 193105482-193105482 on Build GRCh38
rs202181914 Chr.1: 193105629-193105629 on Build GRCh38

View Full Product Details