Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131276900-131277141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
UniGene
Hs.22111
Gene Chromosome Location
Chr.X: 131273506-131289457 on Build GRCh38
Transcripts
NM_001170961.1 NM_001170962.1 NM_001555.4 XM_011531330.1 XM_011531333.1 XM_011531334.1

SNP Detail

SNP ID Chromosome Location
rs751345380 Chr.X: 131277053-131277053 on Build GRCh38
rs761511959 Chr.X: 131276942-131276942 on Build GRCh38
rs376887010 Chr.X: 131277067-131277067 on Build GRCh38
rs747906302 Chr.X: 131277062-131277062 on Build GRCh38
rs757311761 Chr.X: 131277012-131277012 on Build GRCh38
rs779870876 Chr.X: 131277013-131277013 on Build GRCh38
rs1050564340 Chr.X: 131277063-131277063 on Build GRCh38
rs771739503 Chr.X: 131277064-131277064 on Build GRCh38
rs767132682 Chr.X: 131276949-131276949 on Build GRCh38
rs4830219 Chr.X: 131276991-131276991 on Build GRCh38
rs374589007 Chr.X: 131276998-131276998 on Build GRCh38
rs753100283 Chr.X: 131276993-131276993 on Build GRCh38
rs267606355 Chr.X: 131277034-131277034 on Build GRCh38
rs923034176 Chr.X: 131277026-131277026 on Build GRCh38
rs757071351 Chr.X: 131277090-131277090 on Build GRCh38
rs766815289 Chr.X: 131276979-131276979 on Build GRCh38
rs772868171 Chr.X: 131277066-131277066 on Build GRCh38
rs778114359 Chr.X: 131277004-131277004 on Build GRCh38
rs749012041 Chr.X: 131276926-131276926 on Build GRCh38
rs775459929 Chr.X: 131276927-131276927 on Build GRCh38
rs368932840 Chr.X: 131276964-131276964 on Build GRCh38
rs755298160 Chr.X: 131276989-131276989 on Build GRCh38
rs764664783 Chr.X: 131276930-131276930 on Build GRCh38
rs397514622 Chr.X: 131276974-131276974 on Build GRCh38
rs747091505 Chr.X: 131277005-131277005 on Build GRCh38
rs776004054 Chr.X: 131277108-131277108 on Build GRCh38
rs754309168 Chr.X: 131276981-131276981 on Build GRCh38
rs371536225 Chr.X: 131276994-131276994 on Build GRCh38
rs763692298 Chr.X: 131277033-131277033 on Build GRCh38
rs765844658 Chr.X: 131277087-131277087 on Build GRCh38
rs774178023 Chr.X: 131277057-131277057 on Build GRCh38
rs772636261 Chr.X: 131276963-131276963 on Build GRCh38
rs762679632 Chr.X: 131276929-131276929 on Build GRCh38

View Full Product Details