Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99420699-99420926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT1
Entrez Gene ID
2805
Gene Name
glutamic-oxaloacetic transaminase 1
Gene Aliases
AST1, ASTQTL1, GIG18, cAspAT, cCAT
UniGene
Hs.500756
Gene Chromosome Location
Chr.10: 99396870-99430773 on Build GRCh38
Transcripts
NM_002079.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913761 c.149T>C p.V50A Chr.10: 99420775-99420775 on Build GRCh38
6315306 c.198C>A p.S66R Chr.10: 99420726-99420726 on Build GRCh38
244738 c.129G>A p.T43T Chr.10: 99420795-99420795 on Build GRCh38
1160943 c.124C>T p.R42C Chr.10: 99420800-99420800 on Build GRCh38
4010924 c.126C>T p.R42R Chr.10: 99420798-99420798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201941343 Chr.10: 99420744-99420744 on Build GRCh38
rs770053769 Chr.10: 99420735-99420735 on Build GRCh38
rs751629331 Chr.10: 99420800-99420800 on Build GRCh38
rs1037428593 Chr.10: 99420777-99420777 on Build GRCh38
rs763388794 Chr.10: 99420742-99420742 on Build GRCh38
rs956459826 Chr.10: 99420889-99420889 on Build GRCh38
rs377247241 Chr.10: 99420796-99420796 on Build GRCh38
rs921585311 Chr.10: 99420901-99420901 on Build GRCh38
rs201097805 Chr.10: 99420788-99420788 on Build GRCh38
rs145279669 Chr.10: 99420793-99420793 on Build GRCh38
rs780257154 Chr.10: 99420832-99420832 on Build GRCh38
rs776021980 Chr.10: 99420737-99420737 on Build GRCh38
rs200205632 Chr.10: 99420830-99420830 on Build GRCh38
rs564210099 Chr.10: 99420814-99420814 on Build GRCh38
rs372261060 Chr.10: 99420795-99420795 on Build GRCh38
rs752642850 Chr.10: 99420748-99420748 on Build GRCh38
rs749701249 Chr.10: 99420833-99420833 on Build GRCh38
rs769281205 Chr.10: 99420848-99420848 on Build GRCh38
rs781455347 Chr.10: 99420725-99420725 on Build GRCh38
rs758463896 Chr.10: 99420805-99420806 on Build GRCh38
rs11813054 Chr.10: 99420879-99420879 on Build GRCh38
rs889874279 Chr.10: 99420858-99420858 on Build GRCh38
rs1008308357 Chr.10: 99420878-99420878 on Build GRCh38
rs140518658 Chr.10: 99420784-99420784 on Build GRCh38
rs766116233 Chr.10: 99420753-99420753 on Build GRCh38
rs757346593 Chr.10: 99420809-99420809 on Build GRCh38
rs531562990 Chr.10: 99420826-99420826 on Build GRCh38
rs552098213 Chr.10: 99420799-99420799 on Build GRCh38
rs376034578 Chr.10: 99420752-99420752 on Build GRCh38
rs746191161 Chr.10: 99420730-99420730 on Build GRCh38

View Full Product Details