Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55883696-55883951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC1
Entrez Gene ID
55763
Gene Name
exocyst complex component 1
Gene Aliases
BM-102, SEC3, SEC3L1, SEC3P
UniGene
Hs.269665
Gene Chromosome Location
Chr.4: 55853616-55905078 on Build GRCh38
Transcripts
NM_001024924.1 NM_018261.3 NM_178237.2 XM_005265747.2 XM_005265748.1 XM_005265750.2 XM_017008405.1 XM_017008406.1 XM_017008407.1 XM_017008408.1 XM_017008409.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1056190 c.1261G>T p.E421* Chr.4: 55883859-55883859 on Build GRCh38
1618926 c.1318A>T p.S440C Chr.4: 55883916-55883916 on Build GRCh38
1430251 c.1247G>A p.R416Q Chr.4: 55883845-55883845 on Build GRCh38
3409375 c.1246C>T p.R416* Chr.4: 55883844-55883844 on Build GRCh38
1310149 c.1290G>C p.K430N Chr.4: 55883888-55883888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760902141 Chr.4: 55883868-55883868 on Build GRCh38
rs976597592 Chr.4: 55883733-55883733 on Build GRCh38
rs776792823 Chr.4: 55883880-55883880 on Build GRCh38
rs745428581 Chr.4: 55883778-55883778 on Build GRCh38
rs757051243 Chr.4: 55883916-55883916 on Build GRCh38
rs935264025 Chr.4: 55883810-55883810 on Build GRCh38
rs772172832 Chr.4: 55883804-55883805 on Build GRCh38
rs751347826 Chr.4: 55883891-55883891 on Build GRCh38
rs373380748 Chr.4: 55883840-55883840 on Build GRCh38
rs749531381 Chr.4: 55883828-55883828 on Build GRCh38
rs375752640 Chr.4: 55883907-55883907 on Build GRCh38
rs780607773 Chr.4: 55883790-55883790 on Build GRCh38
rs772224328 Chr.4: 55883849-55883849 on Build GRCh38
rs754562716 Chr.4: 55883903-55883903 on Build GRCh38
rs764049101 Chr.4: 55883885-55883885 on Build GRCh38
rs1053533246 Chr.4: 55883795-55883797 on Build GRCh38
rs769153477 Chr.4: 55883829-55883829 on Build GRCh38
rs749766572 Chr.4: 55883803-55883803 on Build GRCh38
rs769282174 Chr.4: 55883789-55883789 on Build GRCh38
rs568448391 Chr.4: 55883842-55883842 on Build GRCh38
rs906418454 Chr.4: 55883924-55883924 on Build GRCh38
rs762821149 Chr.4: 55883883-55883883 on Build GRCh38
rs766570069 Chr.4: 55883874-55883874 on Build GRCh38
rs892249605 Chr.4: 55883815-55883815 on Build GRCh38
rs748505369 Chr.4: 55883847-55883847 on Build GRCh38
rs773547427 Chr.4: 55883866-55883866 on Build GRCh38
rs867077748 Chr.4: 55883844-55883844 on Build GRCh38

View Full Product Details