Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10171889-10172158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLR1
Entrez Gene ID
4973
Gene Name
oxidized low density lipoprotein receptor 1
Gene Aliases
CLEC8A, LOX1, LOXIN, SCARE1, SLOX1
UniGene
Hs.412484
Gene Chromosome Location
Chr.12: 10158300-10172191 on Build GRCh38
Transcripts
NM_001172632.1 NM_001172633.1 NM_002543.3 NM_001079815.1

Gene Symbol
TMEM52B
Entrez Gene ID
120939
Gene Name
transmembrane protein 52B
Gene Aliases
C12orf59
UniGene
Hs.226422
Gene Chromosome Location
Chr.12: 10170599-10191804 on Build GRCh38
Transcripts
NM_001172632.1 NM_001172633.1 NM_002543.3 NM_001079815.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5458322 c.63A>G p.G21G Chr.12: 10172015-10172015 on Build GRCh38
6733320 c.71C>T p.A24V Chr.12: 10172007-10172007 on Build GRCh38
6537607 c.3G>C p.M1I Chr.12: 10172075-10172075 on Build GRCh38
7157396 c.33G>A p.V11V Chr.12: 10172045-10172045 on Build GRCh38
7174062 c.46G>A p.D16N Chr.12: 10172032-10172032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760612923 Chr.12: 10171970-10171970 on Build GRCh38
rs915531617 Chr.12: 10172111-10172111 on Build GRCh38
rs1001170734 Chr.12: 10171934-10171934 on Build GRCh38
rs759750426 Chr.12: 10171980-10171980 on Build GRCh38
rs776662688 Chr.12: 10172105-10172105 on Build GRCh38
rs895561791 Chr.12: 10172007-10172007 on Build GRCh38
rs561719194 Chr.12: 10172088-10172088 on Build GRCh38
rs765382213 Chr.12: 10172119-10172119 on Build GRCh38
rs747863436 Chr.12: 10172075-10172075 on Build GRCh38
rs373714577 Chr.12: 10172087-10172087 on Build GRCh38
rs35462284 Chr.12: 10171926-10171926 on Build GRCh38
rs183354237 Chr.12: 10172062-10172062 on Build GRCh38
rs753085019 Chr.12: 10171999-10171999 on Build GRCh38
rs757079524 Chr.12: 10172106-10172106 on Build GRCh38
rs772986975 Chr.12: 10171965-10171965 on Build GRCh38
rs778379096 Chr.12: 10172072-10172072 on Build GRCh38
rs952517105 Chr.12: 10172051-10172051 on Build GRCh38
rs141892646 Chr.12: 10172019-10172019 on Build GRCh38
rs369592088 Chr.12: 10172102-10172102 on Build GRCh38
rs758642110 Chr.12: 10172005-10172005 on Build GRCh38
rs759662256 Chr.12: 10172117-10172117 on Build GRCh38
rs776752994 Chr.12: 10171974-10171974 on Build GRCh38
rs528823540 Chr.12: 10172110-10172110 on Build GRCh38
rs751951658 Chr.12: 10172034-10172034 on Build GRCh38
rs765417458 Chr.12: 10171984-10171984 on Build GRCh38
rs747703868 Chr.12: 10172009-10172009 on Build GRCh38
rs771699663 Chr.12: 10171941-10171941 on Build GRCh38
rs994354306 Chr.12: 10172014-10172014 on Build GRCh38
rs766392306 Chr.12: 10171973-10171973 on Build GRCh38
rs779706155 Chr.12: 10172046-10172046 on Build GRCh38
rs771920236 Chr.12: 10171958-10171958 on Build GRCh38
rs912071512 Chr.12: 10172098-10172098 on Build GRCh38
rs757897159 Chr.12: 10172035-10172035 on Build GRCh38
rs973122130 Chr.12: 10171976-10171976 on Build GRCh38
rs922622953 Chr.12: 10172126-10172126 on Build GRCh38
rs764522550 Chr.12: 10172029-10172029 on Build GRCh38

View Full Product Details