Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22862978-22863251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB2
Entrez Gene ID
2048
Gene Name
EPH receptor B2
Gene Aliases
CAPB, DRT, EK5, EPHT3, ERK, Hek5, PCBC, Tyro5
UniGene
Hs.380705
Gene Chromosome Location
Chr.1: 22710770-22921500 on Build GRCh38
Transcripts
NM_001309192.1 NM_001309193.1 NM_004442.7 NM_017449.4 XM_006710441.3 XM_006710442.3 NR_036214.1

Gene Symbol
MIR4253
Entrez Gene ID
100422914
Gene Name
microRNA 4253
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 22863159-22863226 on Build GRCh38
Transcripts
NM_001309192.1 NM_001309193.1 NM_004442.7 NM_017449.4 XM_006710441.3 XM_006710442.3 NR_036214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735325 c.890G>A p.C297Y Chr.1: 22863133-22863133 on Build GRCh38
280833 c.853G>A p.A285T Chr.1: 22863078-22863078 on Build GRCh38
131752 c.952G>A p.D318N Chr.1: 22863177-22863177 on Build GRCh38
1205512 c.917G>A p.R306H Chr.1: 22863142-22863142 on Build GRCh38
212329 c.848A>G p.D283G Chr.1: 22863073-22863073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772406361 Chr.1: 22863066-22863066 on Build GRCh38
rs6679387 Chr.1: 22863010-22863010 on Build GRCh38
rs760775609 Chr.1: 22863092-22863092 on Build GRCh38
rs760756497 Chr.1: 22863228-22863228 on Build GRCh38
rs749240821 Chr.1: 22863178-22863178 on Build GRCh38
rs981468344 Chr.1: 22863012-22863012 on Build GRCh38
rs764555559 Chr.1: 22863104-22863104 on Build GRCh38
rs372620832 Chr.1: 22863044-22863044 on Build GRCh38
rs943753954 Chr.1: 22863203-22863203 on Build GRCh38
rs12083517 Chr.1: 22863149-22863149 on Build GRCh38
rs770933694 Chr.1: 22863183-22863183 on Build GRCh38
rs142113032 Chr.1: 22863072-22863072 on Build GRCh38
rs755673470 Chr.1: 22863173-22863173 on Build GRCh38
rs754403229 Chr.1: 22863105-22863105 on Build GRCh38
rs75882465 Chr.1: 22863155-22863155 on Build GRCh38
rs780738757 Chr.1: 22863017-22863017 on Build GRCh38
rs773180070 Chr.1: 22863019-22863019 on Build GRCh38
rs778909458 Chr.1: 22863202-22863202 on Build GRCh38
rs752199354 Chr.1: 22863170-22863170 on Build GRCh38
rs747689340 Chr.1: 22863018-22863018 on Build GRCh38
rs971304652 Chr.1: 22863006-22863006 on Build GRCh38
rs753387416 Chr.1: 22863056-22863056 on Build GRCh38
rs776518179 Chr.1: 22863231-22863231 on Build GRCh38
rs940755901 Chr.1: 22863014-22863014 on Build GRCh38
rs765604107 Chr.1: 22863130-22863130 on Build GRCh38
rs761593747 Chr.1: 22863027-22863028 on Build GRCh38
rs889333439 Chr.1: 22863113-22863113 on Build GRCh38
rs749130502 Chr.1: 22863025-22863025 on Build GRCh38
rs768789658 Chr.1: 22863229-22863229 on Build GRCh38
rs772459312 Chr.1: 22863214-22863214 on Build GRCh38
rs150415344 Chr.1: 22863174-22863174 on Build GRCh38
rs35882952 Chr.1: 22863060-22863060 on Build GRCh38
rs111569109 Chr.1: 22863222-22863222 on Build GRCh38
rs147727992 Chr.1: 22863106-22863106 on Build GRCh38
rs779211309 Chr.1: 22863000-22863000 on Build GRCh38
rs919822792 Chr.1: 22863077-22863077 on Build GRCh38
rs754693296 Chr.1: 22863145-22863145 on Build GRCh38
rs745650276 Chr.1: 22863210-22863210 on Build GRCh38
rs754570824 Chr.1: 22863011-22863011 on Build GRCh38
rs750776222 Chr.1: 22863142-22863142 on Build GRCh38

View Full Product Details