Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158684952-158685189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFDH
Entrez Gene ID
2110
Gene Name
electron transfer flavoprotein dehydrogenase
Gene Aliases
ETFQO, MADD
UniGene
Hs.155729
Gene Chromosome Location
Chr.4: 158672101-158708713 on Build GRCh38
Transcripts
NM_001281737.1 NM_001281738.1 NM_004453.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865975 c.565G>A p.A189T Chr.4: 158685178-158685178 on Build GRCh38
1753612 c.540G>A p.V180V Chr.4: 158685153-158685153 on Build GRCh38
6248082 c.524G>T p.R175L Chr.4: 158685137-158685137 on Build GRCh38
6322684 c.549G>T p.M183I Chr.4: 158685162-158685162 on Build GRCh38
244319 c.525C>G p.R175R Chr.4: 158685138-158685138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980131952 Chr.4: 158685014-158685014 on Build GRCh38
rs773192368 Chr.4: 158685083-158685083 on Build GRCh38
rs973289317 Chr.4: 158685035-158685038 on Build GRCh38
rs535767404 Chr.4: 158685022-158685022 on Build GRCh38
rs375376930 Chr.4: 158685133-158685133 on Build GRCh38
rs767249944 Chr.4: 158685158-158685158 on Build GRCh38
rs759135877 Chr.4: 158685151-158685151 on Build GRCh38
rs755706413 Chr.4: 158685166-158685166 on Build GRCh38
rs752262349 Chr.4: 158685165-158685165 on Build GRCh38
rs190920235 Chr.4: 158685054-158685054 on Build GRCh38
rs775548362 Chr.4: 158685121-158685121 on Build GRCh38
rs121964955 Chr.4: 158685137-158685137 on Build GRCh38
rs747148665 Chr.4: 158685123-158685123 on Build GRCh38
rs377709943 Chr.4: 158685053-158685053 on Build GRCh38
rs999704633 Chr.4: 158685031-158685031 on Build GRCh38
rs774158161 Chr.4: 158685149-158685149 on Build GRCh38
rs901389306 Chr.4: 158685023-158685023 on Build GRCh38
rs768856146 Chr.4: 158685130-158685130 on Build GRCh38
rs746105363 Chr.4: 158685095-158685095 on Build GRCh38
rs762928354 Chr.4: 158685136-158685136 on Build GRCh38
rs900152365 Chr.4: 158685147-158685147 on Build GRCh38
rs374190323 Chr.4: 158685071-158685071 on Build GRCh38
rs180889150 Chr.4: 158685089-158685089 on Build GRCh38
rs772286040 Chr.4: 158685120-158685120 on Build GRCh38
rs201931323 Chr.4: 158685052-158685052 on Build GRCh38
rs755558080 Chr.4: 158685058-158685058 on Build GRCh38
rs753002100 Chr.4: 158685002-158685002 on Build GRCh38
rs778534105 Chr.4: 158685064-158685066 on Build GRCh38

View Full Product Details