Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94491411-94491652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3651927 c.5072C>T p.S1691L Chr.8: 94491646-94491646 on Build GRCh38
7430141 c.5081G>C p.R1694T Chr.8: 94491637-94491637 on Build GRCh38
1578289 c.5076C>T p.G1692G Chr.8: 94491642-94491642 on Build GRCh38
3929674 c.5114G>A p.R1705K Chr.8: 94491604-94491604 on Build GRCh38
3996180 c.5073A>G p.S1691S Chr.8: 94491645-94491645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189777416 Chr.8: 94491629-94491629 on Build GRCh38
rs933730520 Chr.8: 94491453-94491453 on Build GRCh38
rs997433191 Chr.8: 94491470-94491470 on Build GRCh38
rs368214509 Chr.8: 94491562-94491562 on Build GRCh38
rs376110612 Chr.8: 94491566-94491566 on Build GRCh38
rs1046822572 Chr.8: 94491502-94491504 on Build GRCh38
rs745569493 Chr.8: 94491593-94491593 on Build GRCh38
rs767283579 Chr.8: 94491564-94491564 on Build GRCh38
rs548646532 Chr.8: 94491542-94491542 on Build GRCh38
rs777796280 Chr.8: 94491602-94491602 on Build GRCh38
rs766887050 Chr.8: 94491555-94491555 on Build GRCh38
rs900033736 Chr.8: 94491442-94491442 on Build GRCh38
rs534967100 Chr.8: 94491518-94491518 on Build GRCh38
rs568758321 Chr.8: 94491586-94491586 on Build GRCh38
rs941043274 Chr.8: 94491549-94491549 on Build GRCh38
rs755162583 Chr.8: 94491553-94491553 on Build GRCh38

View Full Product Details