Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19058167-19058426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
UniGene
Hs.533468
Gene Chromosome Location
Chr.9: 19053137-19102942 on Build GRCh38
Transcripts
NM_001270890.1 NM_017645.4 XM_011517935.2 XM_017014830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149337 c.2437C>G p.L813V Chr.9: 19058330-19058330 on Build GRCh38
1700975 c.2375C>T p.S792F Chr.9: 19058392-19058392 on Build GRCh38
4402172 c.2478A>G p.S826S Chr.9: 19058289-19058289 on Build GRCh38
3091374 c.2487T>C p.N829N Chr.9: 19058280-19058280 on Build GRCh38
7401209 c.2408C>G p.P803R Chr.9: 19058359-19058359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775826721 Chr.9: 19058317-19058319 on Build GRCh38
rs770193757 Chr.9: 19058304-19058305 on Build GRCh38
rs746397608 Chr.9: 19058211-19058211 on Build GRCh38
rs757894954 Chr.9: 19058357-19058357 on Build GRCh38
rs778585106 Chr.9: 19058197-19058197 on Build GRCh38
rs774757352 Chr.9: 19058225-19058225 on Build GRCh38
rs754485651 Chr.9: 19058252-19058252 on Build GRCh38
rs750186906 Chr.9: 19058237-19058237 on Build GRCh38
rs370946839 Chr.9: 19058329-19058329 on Build GRCh38
rs145576393 Chr.9: 19058256-19058256 on Build GRCh38
rs763005651 Chr.9: 19058215-19058215 on Build GRCh38
rs756480163 Chr.9: 19058374-19058374 on Build GRCh38
rs753790987 Chr.9: 19058241-19058241 on Build GRCh38
rs140508604 Chr.9: 19058389-19058389 on Build GRCh38
rs371362977 Chr.9: 19058232-19058232 on Build GRCh38
rs561181097 Chr.9: 19058379-19058379 on Build GRCh38
rs202204478 Chr.9: 19058218-19058218 on Build GRCh38
rs770185012 Chr.9: 19058275-19058275 on Build GRCh38
rs376889777 Chr.9: 19058266-19058266 on Build GRCh38
rs764215815 Chr.9: 19058223-19058223 on Build GRCh38
rs145639916 Chr.9: 19058398-19058398 on Build GRCh38
rs773565485 Chr.9: 19058315-19058315 on Build GRCh38
rs753748231 Chr.9: 19058322-19058322 on Build GRCh38
rs777456054 Chr.9: 19058365-19058365 on Build GRCh38
rs146530083 Chr.9: 19058366-19058366 on Build GRCh38
rs1039936930 Chr.9: 19058220-19058220 on Build GRCh38
rs781537353 Chr.9: 19058267-19058267 on Build GRCh38
rs201292604 Chr.9: 19058213-19058213 on Build GRCh38
rs747767076 Chr.9: 19058200-19058200 on Build GRCh38
rs759679187 Chr.9: 19058302-19058305 on Build GRCh38
rs1021704117 Chr.9: 19058233-19058233 on Build GRCh38
rs370568570 Chr.9: 19058272-19058272 on Build GRCh38
rs749784720 Chr.9: 19058380-19058380 on Build GRCh38
rs774526121 Chr.9: 19058283-19058283 on Build GRCh38
rs368360263 Chr.9: 19058304-19058304 on Build GRCh38
rs992510729 Chr.9: 19058260-19058260 on Build GRCh38
rs764737576 Chr.9: 19058332-19058332 on Build GRCh38
rs542826832 Chr.9: 19058356-19058356 on Build GRCh38
rs200177865 Chr.9: 19058280-19058280 on Build GRCh38
rs376425978 Chr.9: 19058205-19058205 on Build GRCh38
rs919556447 Chr.9: 19058382-19058382 on Build GRCh38
rs748306154 Chr.9: 19058392-19058392 on Build GRCh38
rs149710112 Chr.9: 19058206-19058206 on Build GRCh38
rs201053056 Chr.9: 19058228-19058228 on Build GRCh38
rs375476349 Chr.9: 19058393-19058393 on Build GRCh38
rs148883960 Chr.9: 19058276-19058276 on Build GRCh38
rs762141378 Chr.9: 19058297-19058297 on Build GRCh38
rs372627388 Chr.9: 19058240-19058240 on Build GRCh38
rs142674219 Chr.9: 19058318-19058318 on Build GRCh38
rs763198448 Chr.9: 19058374-19058374 on Build GRCh38

View Full Product Details