Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35737204-35737471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA2
Entrez Gene ID
57704
Gene Name
glucosylceramidase beta 2
Gene Aliases
AD035, NLGase, SPG46
UniGene
Hs.443134
Gene Chromosome Location
Chr.9: 35736862-35749255 on Build GRCh38
Transcripts
NM_020944.2 XM_005251526.4 XM_006716809.3 XM_011517973.2 XM_017014937.1 XM_017014938.1 XM_017014939.1 XM_017014940.1 XM_017014941.1 XM_017014942.1 XM_017014943.1 XM_017014944.1 XM_017014945.1 XM_017014946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1187718 c.2590G>A p.E864K Chr.9: 35737363-35737363 on Build GRCh38
6234282 c.2581C>A p.Q861K Chr.9: 35737372-35737372 on Build GRCh38
7085157 c.2553C>A p.T851T Chr.9: 35737400-35737400 on Build GRCh38
226217 c.2513C>T p.T838I Chr.9: 35737440-35737440 on Build GRCh38
2774618 c.2549G>A p.R850H Chr.9: 35737404-35737404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778915506 Chr.9: 35737338-35737338 on Build GRCh38
rs772222858 Chr.9: 35737342-35737342 on Build GRCh38
rs758931760 Chr.9: 35737399-35737399 on Build GRCh38
rs527889820 Chr.9: 35737370-35737370 on Build GRCh38
rs777597003 Chr.9: 35737316-35737316 on Build GRCh38
rs374335041 Chr.9: 35737390-35737390 on Build GRCh38
rs762268140 Chr.9: 35737389-35737389 on Build GRCh38
rs764591465 Chr.9: 35737233-35737233 on Build GRCh38
rs765980132 Chr.9: 35737240-35737240 on Build GRCh38
rs780570830 Chr.9: 35737438-35737439 on Build GRCh38
rs145866042 Chr.9: 35737379-35737379 on Build GRCh38
rs752428620 Chr.9: 35737291-35737291 on Build GRCh38
rs752274637 Chr.9: 35737405-35737405 on Build GRCh38
rs1044431904 Chr.9: 35737382-35737382 on Build GRCh38
rs757252849 Chr.9: 35737336-35737336 on Build GRCh38
rs377732636 Chr.9: 35737402-35737402 on Build GRCh38
rs746258856 Chr.9: 35737227-35737228 on Build GRCh38
rs763873057 Chr.9: 35737418-35737418 on Build GRCh38
rs887803349 Chr.9: 35737349-35737349 on Build GRCh38
rs369511568 Chr.9: 35737356-35737356 on Build GRCh38
rs775542106 Chr.9: 35737343-35737343 on Build GRCh38
rs745879966 Chr.9: 35737340-35737340 on Build GRCh38
rs377440003 Chr.9: 35737290-35737290 on Build GRCh38
rs755875971 Chr.9: 35737312-35737312 on Build GRCh38
rs925121854 Chr.9: 35737322-35737322 on Build GRCh38
rs777255160 Chr.9: 35737235-35737235 on Build GRCh38
rs754527190 Chr.9: 35737251-35737251 on Build GRCh38
rs762468037 Chr.9: 35737236-35737236 on Build GRCh38
rs892032709 Chr.9: 35737417-35737417 on Build GRCh38
rs199510198 Chr.9: 35737344-35737344 on Build GRCh38
rs191410452 Chr.9: 35737398-35737398 on Build GRCh38
rs1012890540 Chr.9: 35737373-35737373 on Build GRCh38
rs749211700 Chr.9: 35737318-35737318 on Build GRCh38
rs398123015 Chr.9: 35737335-35737335 on Build GRCh38
rs912331543 Chr.9: 35737252-35737252 on Build GRCh38
rs760907481 Chr.9: 35737224-35737224 on Build GRCh38
rs918460586 Chr.9: 35737294-35737294 on Build GRCh38
rs756367976 Chr.9: 35737238-35737239 on Build GRCh38
rs138519801 Chr.9: 35737413-35737413 on Build GRCh38
rs767031907 Chr.9: 35737254-35737254 on Build GRCh38

View Full Product Details