Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33576104-33576347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS12
Entrez Gene ID
81792
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 12
Gene Aliases
PRO4389
UniGene
Hs.12680
Gene Chromosome Location
Chr.5: 33523535-33892180 on Build GRCh38
Transcripts
NM_001324512.1 NM_030955.3 XM_017009905.1 XM_017009906.1 XM_017009907.1 XM_017009908.1 XM_017009909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354255 c.3704G>A p.R1235K Chr.5: 33576322-33576322 on Build GRCh38
738083 c.3804C>A p.H1268Q Chr.5: 33576222-33576222 on Build GRCh38
3015015 c.3785A>T p.K1262M Chr.5: 33576241-33576241 on Build GRCh38
4829724 c.3772G>C p.E1258Q Chr.5: 33576254-33576254 on Build GRCh38
301634 c.3721A>G p.T1241A Chr.5: 33576305-33576305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145214580 Chr.5: 33576305-33576305 on Build GRCh38
rs762255827 Chr.5: 33576272-33576272 on Build GRCh38
rs763382391 Chr.5: 33576193-33576193 on Build GRCh38
rs749342789 Chr.5: 33576237-33576237 on Build GRCh38
rs370406310 Chr.5: 33576204-33576204 on Build GRCh38
rs756351049 Chr.5: 33576230-33576230 on Build GRCh38
rs976612104 Chr.5: 33576239-33576239 on Build GRCh38
rs35581318 Chr.5: 33576134-33576134 on Build GRCh38
rs373062449 Chr.5: 33576289-33576289 on Build GRCh38
rs766859897 Chr.5: 33576195-33576195 on Build GRCh38
rs911174816 Chr.5: 33576222-33576222 on Build GRCh38
rs929760262 Chr.5: 33576288-33576288 on Build GRCh38
rs777715999 Chr.5: 33576234-33576234 on Build GRCh38
rs201193243 Chr.5: 33576252-33576252 on Build GRCh38
rs774040470 Chr.5: 33576129-33576129 on Build GRCh38
rs772051667 Chr.5: 33576280-33576280 on Build GRCh38
rs757124483 Chr.5: 33576283-33576284 on Build GRCh38
rs771965526 Chr.5: 33576150-33576150 on Build GRCh38
rs373958116 Chr.5: 33576215-33576215 on Build GRCh38
rs765913018 Chr.5: 33576287-33576287 on Build GRCh38
rs753978117 Chr.5: 33576317-33576317 on Build GRCh38
rs774886665 Chr.5: 33576302-33576302 on Build GRCh38
rs199947257 Chr.5: 33576297-33576297 on Build GRCh38
rs189970267 Chr.5: 33576296-33576296 on Build GRCh38
rs760821924 Chr.5: 33576264-33576264 on Build GRCh38
rs764620883 Chr.5: 33576301-33576301 on Build GRCh38
rs767934370 Chr.5: 33576309-33576309 on Build GRCh38
rs746646285 Chr.5: 33576291-33576291 on Build GRCh38
rs200655889 Chr.5: 33576324-33576324 on Build GRCh38
rs757177041 Chr.5: 33576238-33576238 on Build GRCh38
rs753323743 Chr.5: 33576184-33576184 on Build GRCh38
rs760392962 Chr.5: 33576163-33576163 on Build GRCh38
rs922399712 Chr.5: 33576263-33576263 on Build GRCh38
rs756316670 Chr.5: 33576312-33576312 on Build GRCh38
rs751950320 Chr.5: 33576203-33576203 on Build GRCh38
rs752989182 Chr.5: 33576310-33576310 on Build GRCh38
rs764340709 Chr.5: 33576314-33576314 on Build GRCh38
rs550863790 Chr.5: 33576144-33576144 on Build GRCh38
rs958239595 Chr.5: 33576177-33576177 on Build GRCh38
rs776876572 Chr.5: 33576254-33576254 on Build GRCh38
rs752972657 Chr.5: 33576229-33576229 on Build GRCh38
rs775332386 Chr.5: 33576151-33576151 on Build GRCh38
rs763578276 Chr.5: 33576171-33576171 on Build GRCh38
rs572898982 Chr.5: 33576320-33576320 on Build GRCh38

View Full Product Details