Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35954313-35954578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNBD2
Entrez Gene ID
140894
Gene Name
cyclic nucleotide binding domain containing 2
Gene Aliases
C20orf152, CNMPD1
UniGene
Hs.732697
Gene Chromosome Location
Chr.20: 35954561-36030700 on Build GRCh38
Transcripts
XM_011528590.2 XM_011528593.2 NM_016558.3 NM_033630.2

Gene Symbol
SCAND1
Entrez Gene ID
51282
Gene Name
SCAN domain containing 1
Gene Aliases
RAZ1, SDP1
UniGene
Hs.584909
Gene Chromosome Location
Chr.20: 35953617-35955359 on Build GRCh38
Transcripts
XM_011528590.2 XM_011528593.2 NM_016558.3 NM_033630.2

SNP Detail

SNP ID Chromosome Location
rs755111676 Chr.20: 35954480-35954480 on Build GRCh38
rs776076848 Chr.20: 35954338-35954338 on Build GRCh38
rs961966738 Chr.20: 35954436-35954436 on Build GRCh38
rs766030962 Chr.20: 35954360-35954360 on Build GRCh38
rs567963384 Chr.20: 35954403-35954403 on Build GRCh38
rs761280979 Chr.20: 35954342-35954342 on Build GRCh38
rs781246551 Chr.20: 35954364-35954364 on Build GRCh38
rs901140881 Chr.20: 35954481-35954481 on Build GRCh38
rs549558169 Chr.20: 35954397-35954397 on Build GRCh38
rs1049569361 Chr.20: 35954527-35954527 on Build GRCh38
rs34688876 Chr.20: 35954532-35954532 on Build GRCh38
rs35562811 Chr.20: 35954482-35954482 on Build GRCh38
rs992274724 Chr.20: 35954454-35954454 on Build GRCh38
rs549781220 Chr.20: 35954420-35954420 on Build GRCh38
rs35005711 Chr.20: 35954534-35954534 on Build GRCh38
rs770401022 Chr.20: 35954511-35954511 on Build GRCh38
rs748060735 Chr.20: 35954346-35954346 on Build GRCh38
rs1043629208 Chr.20: 35954391-35954391 on Build GRCh38
rs751396367 Chr.20: 35954361-35954361 on Build GRCh38
rs113606286 Chr.20: 35954395-35954395 on Build GRCh38
rs1053110706 Chr.20: 35954421-35954421 on Build GRCh38
rs772464390 Chr.20: 35954399-35954399 on Build GRCh38
rs575970060 Chr.20: 35954453-35954453 on Build GRCh38
rs769228919 Chr.20: 35954412-35954412 on Build GRCh38
rs771989900 Chr.20: 35954384-35954385 on Build GRCh38
rs41293096 Chr.20: 35954429-35954429 on Build GRCh38
rs748148891 Chr.20: 35954368-35954368 on Build GRCh38
rs750073966 Chr.20: 35954347-35954347 on Build GRCh38
rs993059511 Chr.20: 35954456-35954456 on Build GRCh38
rs746125345 Chr.20: 35954469-35954469 on Build GRCh38
rs980211134 Chr.20: 35954444-35954444 on Build GRCh38
rs35448195 Chr.20: 35954443-35954443 on Build GRCh38
rs35694954 Chr.20: 35954438-35954438 on Build GRCh38
rs779210895 Chr.20: 35954549-35954549 on Build GRCh38
rs58346765 Chr.20: 35954378-35954378 on Build GRCh38
rs891243566 Chr.20: 35954531-35954531 on Build GRCh38
rs1028123672 Chr.20: 35954434-35954434 on Build GRCh38
rs896916941 Chr.20: 35954540-35954540 on Build GRCh38
rs527906457 Chr.20: 35954386-35954386 on Build GRCh38
rs551565066 Chr.20: 35954356-35954356 on Build GRCh38
rs749400317 Chr.20: 35954376-35954376 on Build GRCh38
rs556551814 Chr.20: 35954486-35954486 on Build GRCh38
rs3180131 Chr.20: 35954465-35954465 on Build GRCh38
rs762497207 Chr.20: 35954488-35954488 on Build GRCh38
rs756246369 Chr.20: 35954370-35954370 on Build GRCh38
rs554337530 Chr.20: 35954452-35954452 on Build GRCh38
rs146120217 Chr.20: 35954442-35954442 on Build GRCh38
rs751215098 Chr.20: 35954425-35954425 on Build GRCh38
rs961099254 Chr.20: 35954445-35954445 on Build GRCh38
rs778063491 Chr.20: 35954375-35954375 on Build GRCh38
rs779165480 Chr.20: 35954468-35954468 on Build GRCh38
rs772723312 Chr.20: 35954415-35954415 on Build GRCh38
rs762818886 Chr.20: 35954411-35954411 on Build GRCh38
rs545354984 Chr.20: 35954542-35954542 on Build GRCh38
rs1016695599 Chr.20: 35954497-35954497 on Build GRCh38
rs759356596 Chr.20: 35954428-35954428 on Build GRCh38
rs193293635 Chr.20: 35954448-35954448 on Build GRCh38
rs41293094 Chr.20: 35954346-35954346 on Build GRCh38
rs532041761 Chr.20: 35954419-35954419 on Build GRCh38
rs772423352 Chr.20: 35954470-35954470 on Build GRCh38

View Full Product Details