Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229518326-229518551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB10
Entrez Gene ID
23456
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
EST20237, M-ABC2, MTABC2
UniGene
Hs.17614
Gene Chromosome Location
Chr.1: 229516582-229558987 on Build GRCh38
Transcripts
NM_012089.2 XM_011544135.2 XM_011544136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6541941 c.2048G>A p.R683K Chr.1: 229518348-229518348 on Build GRCh38
4400020 c.2060T>G p.V687G Chr.1: 229518336-229518336 on Build GRCh38
6326077 c.2017C>T p.Q673* Chr.1: 229518379-229518379 on Build GRCh38
5574368 c.2031_2034delTCGA p.R678fs*1 Chr.1: 229518362-229518365 on Build GRCh38
6869630 c.1999G>T p.E667* Chr.1: 229518397-229518397 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs970122557 Chr.1: 229518357-229518358 on Build GRCh38
rs751596334 Chr.1: 229518407-229518407 on Build GRCh38
rs755055634 Chr.1: 229518409-229518409 on Build GRCh38
rs141486923 Chr.1: 229518396-229518396 on Build GRCh38
rs373579061 Chr.1: 229518373-229518373 on Build GRCh38
rs201773661 Chr.1: 229518351-229518351 on Build GRCh38
rs563279613 Chr.1: 229518474-229518474 on Build GRCh38
rs940606886 Chr.1: 229518473-229518476 on Build GRCh38
rs371157444 Chr.1: 229518445-229518445 on Build GRCh38
rs773072766 Chr.1: 229518398-229518398 on Build GRCh38
rs780813956 Chr.1: 229518397-229518397 on Build GRCh38
rs746437184 Chr.1: 229518456-229518456 on Build GRCh38
rs770623136 Chr.1: 229518354-229518354 on Build GRCh38
rs776339083 Chr.1: 229518390-229518390 on Build GRCh38
rs376233356 Chr.1: 229518446-229518446 on Build GRCh38
rs752952060 Chr.1: 229518418-229518418 on Build GRCh38
rs898976203 Chr.1: 229518508-229518508 on Build GRCh38
rs185229946 Chr.1: 229518363-229518363 on Build GRCh38
rs762870986 Chr.1: 229518399-229518399 on Build GRCh38
rs768196942 Chr.1: 229518461-229518461 on Build GRCh38
rs377675121 Chr.1: 229518443-229518443 on Build GRCh38
rs767561420 Chr.1: 229518416-229518416 on Build GRCh38
rs999114812 Chr.1: 229518482-229518482 on Build GRCh38
rs150786524 Chr.1: 229518441-229518441 on Build GRCh38
rs756235149 Chr.1: 229518423-229518423 on Build GRCh38
rs766368971 Chr.1: 229518406-229518406 on Build GRCh38
rs995092599 Chr.1: 229518509-229518509 on Build GRCh38
rs777979470 Chr.1: 229518426-229518426 on Build GRCh38
rs367565268 Chr.1: 229518450-229518450 on Build GRCh38

View Full Product Details