Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9068659-9068894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2M
Entrez Gene ID
2
Gene Name
alpha-2-macroglobulin
Gene Aliases
A2MD, CPAMD5, FWP007, S863-7
UniGene
Hs.212838
Gene Chromosome Location
Chr.12: 9067708-9115962 on Build GRCh38
Transcripts
NM_000014.4 XM_006719056.2 XM_017018683.1 XM_017018684.1 XM_017018685.1

Gene Symbol
KLRG1
Entrez Gene ID
10219
Gene Name
killer cell lectin like receptor G1
Gene Aliases
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE
UniGene
Hs.558446
Gene Chromosome Location
Chr.12: 8989079-9255692 on Build GRCh38
Transcripts
NM_000014.4 XM_006719056.2 XM_017018683.1 XM_017018684.1 XM_017018685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424984 c.4364C>T p.T1455M Chr.12: 9068742-9068742 on Build GRCh38
695462 c.4264-2A>G p.? Chr.12: 9068844-9068844 on Build GRCh38
4891318 c.4336A>G p.I1446V Chr.12: 9068770-9068770 on Build GRCh38
6888246 c.4318A>G p.R1440G Chr.12: 9068788-9068788 on Build GRCh38
2045014 c.4359C>T p.Y1453Y Chr.12: 9068747-9068747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768638522 Chr.12: 9068759-9068759 on Build GRCh38
rs201115819 Chr.12: 9068811-9068811 on Build GRCh38
rs976898073 Chr.12: 9068798-9068798 on Build GRCh38
rs761933862 Chr.12: 9068782-9068782 on Build GRCh38
rs745544071 Chr.12: 9068744-9068744 on Build GRCh38
rs746496048 Chr.12: 9068733-9068733 on Build GRCh38
rs775058537 Chr.12: 9068731-9068734 on Build GRCh38
rs1802965 Chr.12: 9068815-9068815 on Build GRCh38
rs775436209 Chr.12: 9068756-9068756 on Build GRCh38
rs111819233 Chr.12: 9068739-9068739 on Build GRCh38
rs188015653 Chr.12: 9068810-9068810 on Build GRCh38
rs370110696 Chr.12: 9068719-9068719 on Build GRCh38
rs779354961 Chr.12: 9068726-9068726 on Build GRCh38
rs977089869 Chr.12: 9068827-9068827 on Build GRCh38
rs774199694 Chr.12: 9068777-9068777 on Build GRCh38
rs766672959 Chr.12: 9068833-9068833 on Build GRCh38
rs749692631 Chr.12: 9068718-9068718 on Build GRCh38
rs1017680609 Chr.12: 9068684-9068684 on Build GRCh38
rs369658444 Chr.12: 9068747-9068747 on Build GRCh38
rs965842592 Chr.12: 9068792-9068792 on Build GRCh38
rs376343602 Chr.12: 9068742-9068742 on Build GRCh38
rs754217279 Chr.12: 9068842-9068842 on Build GRCh38
rs201382604 Chr.12: 9068757-9068757 on Build GRCh38
rs767608990 Chr.12: 9068803-9068803 on Build GRCh38

View Full Product Details