Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9490988-9491261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM17
Entrez Gene ID
6868
Gene Name
ADAM metallopeptidase domain 17
Gene Aliases
ADAM18, CD156B, CSVP, NISBD, NISBD1, TACE
UniGene
Hs.404914
Gene Chromosome Location
Chr.2: 9488486-9555820 on Build GRCh38
Transcripts
NM_003183.5 XM_011510375.2 XM_011510376.2 XM_011510377.2 XM_017004785.1 XM_017004786.1

Gene Symbol
IAH1
Entrez Gene ID
285148
Gene Name
isoamyl acetate-hydrolyzing esterase 1 homolog
Gene Aliases
-
UniGene
Hs.667226
Gene Chromosome Location
Chr.2: 9473638-9512403 on Build GRCh38
Transcripts
NM_003183.5 XM_011510375.2 XM_011510376.2 XM_011510377.2 XM_017004785.1 XM_017004786.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6289631 c.2083-6T>C p.? Chr.2: 9491157-9491157 on Build GRCh38
4805964 c.2084A>T p.D695V Chr.2: 9491150-9491150 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760627744 Chr.2: 9491094-9491094 on Build GRCh38
rs781600390 Chr.2: 9491178-9491178 on Build GRCh38
rs574369077 Chr.2: 9491016-9491016 on Build GRCh38
rs776074945 Chr.2: 9491065-9491065 on Build GRCh38
rs746987344 Chr.2: 9491201-9491201 on Build GRCh38
rs755493332 Chr.2: 9491174-9491174 on Build GRCh38
rs907060873 Chr.2: 9491205-9491205 on Build GRCh38
rs756149983 Chr.2: 9491182-9491187 on Build GRCh38
rs372459687 Chr.2: 9491160-9491160 on Build GRCh38
rs945189134 Chr.2: 9491042-9491042 on Build GRCh38
rs371300216 Chr.2: 9491190-9491190 on Build GRCh38
rs770956800 Chr.2: 9491191-9491191 on Build GRCh38
rs372419048 Chr.2: 9491063-9491063 on Build GRCh38
rs758135029 Chr.2: 9491200-9491203 on Build GRCh38
rs543077790 Chr.2: 9491043-9491043 on Build GRCh38
rs903700740 Chr.2: 9491093-9491093 on Build GRCh38
rs752991112 Chr.2: 9491179-9491179 on Build GRCh38
rs778028713 Chr.2: 9491109-9491109 on Build GRCh38
rs576343468 Chr.2: 9491142-9491142 on Build GRCh38
rs764926423 Chr.2: 9491119-9491119 on Build GRCh38
rs767985722 Chr.2: 9491120-9491121 on Build GRCh38
rs747163792 Chr.2: 9491123-9491123 on Build GRCh38
rs765883520 Chr.2: 9491137-9491137 on Build GRCh38
rs751870775 Chr.2: 9491074-9491075 on Build GRCh38
rs766698101 Chr.2: 9491068-9491068 on Build GRCh38
rs776564382 Chr.2: 9491115-9491115 on Build GRCh38
rs760143401 Chr.2: 9491114-9491115 on Build GRCh38
rs189136851 Chr.2: 9491067-9491067 on Build GRCh38
rs769997193 Chr.2: 9491062-9491062 on Build GRCh38
rs756339806 Chr.2: 9491182-9491182 on Build GRCh38
rs755646768 Chr.2: 9491195-9491196 on Build GRCh38
rs370157347 Chr.2: 9491116-9491116 on Build GRCh38
rs768383645 Chr.2: 9491107-9491107 on Build GRCh38
rs749976457 Chr.2: 9491127-9491127 on Build GRCh38
rs772069221 Chr.2: 9491068-9491068 on Build GRCh38
rs1055146141 Chr.2: 9491208-9491208 on Build GRCh38
rs779027718 Chr.2: 9491192-9491192 on Build GRCh38
rs1038388537 Chr.2: 9491113-9491113 on Build GRCh38
rs10495562 Chr.2: 9491211-9491211 on Build GRCh38
rs770292932 Chr.2: 9491022-9491022 on Build GRCh38
rs777922224 Chr.2: 9491183-9491183 on Build GRCh38
rs764325373 Chr.2: 9491192-9491193 on Build GRCh38
rs377163769 Chr.2: 9491088-9491088 on Build GRCh38
rs762334638 Chr.2: 9491130-9491130 on Build GRCh38
rs1050032598 Chr.2: 9491032-9491032 on Build GRCh38
rs994701272 Chr.2: 9491163-9491163 on Build GRCh38
rs763238472 Chr.2: 9491052-9491052 on Build GRCh38

View Full Product Details