Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77293047-77293302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
UniGene
Hs.188746
Gene Chromosome Location
Chr.16: 77282128-77435114 on Build GRCh38
Transcripts
NM_001326358.1 NM_199355.3 XM_011522923.1 XM_011522924.1 XM_017022988.1 XM_017022989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653359 c.3147C>T p.P1049P Chr.16: 77293118-77293118 on Build GRCh38
5481326 c.3082G>A p.E1028K Chr.16: 77293183-77293183 on Build GRCh38
3512486 c.3045A>G p.E1015E Chr.16: 77293220-77293220 on Build GRCh38
973896 c.3165G>A p.Q1055Q Chr.16: 77293100-77293100 on Build GRCh38
4429163 c.3084G>C p.E1028D Chr.16: 77293181-77293181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764081033 Chr.16: 77293160-77293160 on Build GRCh38
rs906746417 Chr.16: 77293244-77293244 on Build GRCh38
rs778665951 Chr.16: 77293254-77293254 on Build GRCh38
rs1023239338 Chr.16: 77293170-77293170 on Build GRCh38
rs769604667 Chr.16: 77293112-77293112 on Build GRCh38
rs201570714 Chr.16: 77293259-77293259 on Build GRCh38
rs981963321 Chr.16: 77293178-77293178 on Build GRCh38
rs766138350 Chr.16: 77293142-77293142 on Build GRCh38
rs376264920 Chr.16: 77293196-77293196 on Build GRCh38
rs377705872 Chr.16: 77293166-77293166 on Build GRCh38
rs370743740 Chr.16: 77293198-77293198 on Build GRCh38
rs754618023 Chr.16: 77293253-77293253 on Build GRCh38
rs112547949 Chr.16: 77293093-77293093 on Build GRCh38
rs200998751 Chr.16: 77293264-77293264 on Build GRCh38
rs374112306 Chr.16: 77293199-77293199 on Build GRCh38
rs893547958 Chr.16: 77293128-77293128 on Build GRCh38
rs754174411 Chr.16: 77293248-77293248 on Build GRCh38
rs1021968711 Chr.16: 77293127-77293127 on Build GRCh38
rs760193459 Chr.16: 77293223-77293224 on Build GRCh38
rs776339298 Chr.16: 77293234-77293234 on Build GRCh38
rs758639413 Chr.16: 77293148-77293148 on Build GRCh38
rs752883058 Chr.16: 77293079-77293079 on Build GRCh38
rs756685685 Chr.16: 77293241-77293241 on Build GRCh38
rs771451726 Chr.16: 77293216-77293216 on Build GRCh38
rs561262222 Chr.16: 77293119-77293119 on Build GRCh38
rs751029682 Chr.16: 77293240-77293240 on Build GRCh38
rs145095974 Chr.16: 77293237-77293237 on Build GRCh38
rs780573259 Chr.16: 77293247-77293247 on Build GRCh38
rs1011337681 Chr.16: 77293271-77293271 on Build GRCh38
rs761662175 Chr.16: 77293123-77293123 on Build GRCh38
rs747733216 Chr.16: 77293257-77293257 on Build GRCh38
rs139701298 Chr.16: 77293224-77293224 on Build GRCh38
rs369716907 Chr.16: 77293116-77293116 on Build GRCh38
rs61754842 Chr.16: 77293145-77293145 on Build GRCh38
rs187998780 Chr.16: 77293094-77293094 on Build GRCh38
rs1011453043 Chr.16: 77293126-77293126 on Build GRCh38
rs1046537063 Chr.16: 77293242-77293242 on Build GRCh38
rs760500555 Chr.16: 77293134-77293134 on Build GRCh38
rs377137286 Chr.16: 77293217-77293217 on Build GRCh38
rs765403489 Chr.16: 77293078-77293078 on Build GRCh38
rs201935168 Chr.16: 77293091-77293091 on Build GRCh38
rs369984540 Chr.16: 77293183-77293183 on Build GRCh38
rs376965912 Chr.16: 77293262-77293262 on Build GRCh38
rs780554901 Chr.16: 77293188-77293188 on Build GRCh38
rs781753083 Chr.16: 77293179-77293179 on Build GRCh38
rs144398542 Chr.16: 77293086-77293086 on Build GRCh38
rs752422457 Chr.16: 77293221-77293222 on Build GRCh38
rs149947117 Chr.16: 77293235-77293235 on Build GRCh38
rs200775058 Chr.16: 77293225-77293225 on Build GRCh38
rs755215391 Chr.16: 77293077-77293077 on Build GRCh38
rs368889887 Chr.16: 77293265-77293265 on Build GRCh38
rs747653054 Chr.16: 77293205-77293205 on Build GRCh38
rs373173918 Chr.16: 77293184-77293184 on Build GRCh38
rs142167857 Chr.16: 77293239-77293239 on Build GRCh38
rs1052613609 Chr.16: 77293272-77293272 on Build GRCh38
rs534278374 Chr.16: 77293107-77293107 on Build GRCh38
rs767283285 Chr.16: 77293070-77293070 on Build GRCh38
rs757730361 Chr.16: 77293173-77293173 on Build GRCh38
rs149632847 Chr.16: 77293085-77293085 on Build GRCh38
rs139644893 Chr.16: 77293125-77293125 on Build GRCh38
rs35296483 Chr.16: 77293121-77293121 on Build GRCh38
rs148703569 Chr.16: 77293108-77293108 on Build GRCh38
rs768111531 Chr.16: 77293120-77293120 on Build GRCh38
rs967174946 Chr.16: 77293133-77293133 on Build GRCh38
rs1049158799 Chr.16: 77293074-77293074 on Build GRCh38
rs1019481262 Chr.16: 77293161-77293161 on Build GRCh38
rs775296189 Chr.16: 77293100-77293100 on Build GRCh38
rs766563853 Chr.16: 77293267-77293267 on Build GRCh38

View Full Product Details