Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 905346-905591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCYAP1
Entrez Gene ID
116
Gene Name
adenylate cyclase activating polypeptide 1
Gene Aliases
PACAP
UniGene
Hs.531719
Gene Chromosome Location
Chr.18: 904342-912172 on Build GRCh38
Transcripts
NM_001099733.1 NM_001117.4 XM_005258081.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
710337 c.15C>A p.S5R Chr.18: 905401-905401 on Build GRCh38
3959441 c.110+4G>T p.? Chr.18: 905500-905500 on Build GRCh38
324648 c.56A>G p.H19R Chr.18: 905442-905442 on Build GRCh38
710336 c.75A>C p.S25S Chr.18: 905461-905461 on Build GRCh38
3422350 c.88G>A p.G30R Chr.18: 905474-905474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533218516 Chr.18: 905572-905573 on Build GRCh38
rs776807884 Chr.18: 905528-905528 on Build GRCh38
rs769265867 Chr.18: 905428-905428 on Build GRCh38
rs916823015 Chr.18: 905469-905469 on Build GRCh38
rs765316211 Chr.18: 905572-905572 on Build GRCh38
rs760474019 Chr.18: 905470-905470 on Build GRCh38
rs774142783 Chr.18: 905451-905451 on Build GRCh38
rs958463338 Chr.18: 905464-905464 on Build GRCh38
rs765029234 Chr.18: 905472-905472 on Build GRCh38
rs989726208 Chr.18: 905467-905467 on Build GRCh38
rs755052187 Chr.18: 905380-905380 on Build GRCh38
rs928341128 Chr.18: 905499-905499 on Build GRCh38
rs781723482 Chr.18: 905542-905542 on Build GRCh38
rs576246004 Chr.18: 905567-905567 on Build GRCh38
rs747342728 Chr.18: 905484-905484 on Build GRCh38
rs906826357 Chr.18: 905399-905399 on Build GRCh38
rs778244814 Chr.18: 905420-905420 on Build GRCh38
rs749007403 Chr.18: 905491-905491 on Build GRCh38
rs765402609 Chr.18: 905477-905477 on Build GRCh38
rs1031162701 Chr.18: 905448-905448 on Build GRCh38
rs757684819 Chr.18: 905538-905538 on Build GRCh38
rs552653569 Chr.18: 905516-905516 on Build GRCh38
rs534614195 Chr.18: 905513-905513 on Build GRCh38
rs758664236 Chr.18: 905416-905416 on Build GRCh38
rs760455583 Chr.18: 905519-905519 on Build GRCh38
rs568852695 Chr.18: 905550-905550 on Build GRCh38
rs370963358 Chr.18: 905530-905530 on Build GRCh38
rs544682110 Chr.18: 905556-905556 on Build GRCh38
rs573823992 Chr.18: 905488-905488 on Build GRCh38
rs753855693 Chr.18: 905368-905368 on Build GRCh38
rs766216691 Chr.18: 905527-905527 on Build GRCh38
rs924669214 Chr.18: 905536-905537 on Build GRCh38
rs373051517 Chr.18: 905474-905474 on Build GRCh38
rs377558347 Chr.18: 905427-905427 on Build GRCh38
rs758901557 Chr.18: 905481-905481 on Build GRCh38
rs1051750118 Chr.18: 905564-905564 on Build GRCh38
rs761860008 Chr.18: 905461-905461 on Build GRCh38
rs747367878 Chr.18: 905423-905423 on Build GRCh38
rs748744145 Chr.18: 905401-905401 on Build GRCh38
rs773026207 Chr.18: 905518-905518 on Build GRCh38
rs771985769 Chr.18: 905517-905517 on Build GRCh38
rs779385024 Chr.18: 905391-905391 on Build GRCh38
rs1002629931 Chr.18: 905414-905414 on Build GRCh38
rs757703280 Chr.18: 905485-905485 on Build GRCh38
rs748742011 Chr.18: 905450-905450 on Build GRCh38
rs778168865 Chr.18: 905482-905482 on Build GRCh38
rs754609134 Chr.18: 905543-905543 on Build GRCh38
rs2231184 Chr.18: 905558-905558 on Build GRCh38
rs752778053 Chr.18: 905533-905533 on Build GRCh38
rs975425690 Chr.18: 905522-905522 on Build GRCh38
rs755035529 Chr.18: 905475-905475 on Build GRCh38
rs773871169 Chr.18: 905515-905515 on Build GRCh38
rs2231183 Chr.18: 905536-905536 on Build GRCh38
rs1037583726 Chr.18: 905547-905547 on Build GRCh38
rs907671106 Chr.18: 905531-905531 on Build GRCh38
rs35305428 Chr.18: 905479-905479 on Build GRCh38
rs8192594 Chr.18: 905468-905468 on Build GRCh38
rs761954441 Chr.18: 905548-905548 on Build GRCh38
rs938413845 Chr.18: 905508-905508 on Build GRCh38
rs766609304 Chr.18: 905365-905365 on Build GRCh38

View Full Product Details