Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29562893-29563118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD15
Entrez Gene ID
116236
Gene Name
abhydrolase domain containing 15
Gene Aliases
-
UniGene
Hs.106510
Gene Chromosome Location
Chr.17: 29560661-29567024 on Build GRCh38
Transcripts
NM_198147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755369 c.1000A>G p.T334A Chr.17: 29562968-29562968 on Build GRCh38
4129799 c.1023G>C p.P341P Chr.17: 29562945-29562945 on Build GRCh38
296802 c.935G>A p.R312H Chr.17: 29563033-29563033 on Build GRCh38
6556261 c.1026C>A p.L342L Chr.17: 29562942-29562942 on Build GRCh38
5423631 c.1003T>G p.Y335D Chr.17: 29562965-29562965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145378805 Chr.17: 29563029-29563029 on Build GRCh38
rs974051668 Chr.17: 29562970-29562970 on Build GRCh38
rs776728486 Chr.17: 29562932-29562932 on Build GRCh38
rs752839323 Chr.17: 29563029-29563030 on Build GRCh38
rs377743368 Chr.17: 29562938-29562938 on Build GRCh38
rs753440125 Chr.17: 29562951-29562951 on Build GRCh38
rs768072108 Chr.17: 29562968-29562969 on Build GRCh38
rs145964335 Chr.17: 29562946-29562946 on Build GRCh38
rs139847076 Chr.17: 29562961-29562961 on Build GRCh38
rs766113718 Chr.17: 29562926-29562926 on Build GRCh38
rs142517096 Chr.17: 29563062-29563062 on Build GRCh38
rs990893896 Chr.17: 29562958-29562958 on Build GRCh38
rs151130735 Chr.17: 29562971-29562971 on Build GRCh38
rs759734185 Chr.17: 29562933-29562933 on Build GRCh38
rs201245024 Chr.17: 29563095-29563095 on Build GRCh38
rs542939 Chr.17: 29562968-29562968 on Build GRCh38
rs760494055 Chr.17: 29562923-29562923 on Build GRCh38
rs200949331 Chr.17: 29562921-29562921 on Build GRCh38
rs960885801 Chr.17: 29563081-29563081 on Build GRCh38
rs764221422 Chr.17: 29563020-29563020 on Build GRCh38
rs754905589 Chr.17: 29563034-29563034 on Build GRCh38
rs370276970 Chr.17: 29562956-29562956 on Build GRCh38
rs150059487 Chr.17: 29563025-29563025 on Build GRCh38
rs752294131 Chr.17: 29563052-29563052 on Build GRCh38
rs721479 Chr.17: 29562945-29562945 on Build GRCh38
rs367783992 Chr.17: 29562988-29562988 on Build GRCh38
rs761726141 Chr.17: 29562942-29562942 on Build GRCh38
rs764412151 Chr.17: 29563037-29563038 on Build GRCh38
rs376067361 Chr.17: 29562994-29562994 on Build GRCh38
rs989211505 Chr.17: 29562999-29562999 on Build GRCh38
rs752129442 Chr.17: 29563024-29563024 on Build GRCh38
rs763156782 Chr.17: 29563009-29563009 on Build GRCh38
rs778655705 Chr.17: 29563045-29563045 on Build GRCh38
rs758533372 Chr.17: 29562941-29562941 on Build GRCh38
rs563663528 Chr.17: 29562987-29562987 on Build GRCh38
rs540488510 Chr.17: 29562955-29562955 on Build GRCh38
rs776555358 Chr.17: 29562984-29562984 on Build GRCh38
rs201536407 Chr.17: 29563033-29563033 on Build GRCh38
rs139299388 Chr.17: 29562940-29562940 on Build GRCh38
rs774839302 Chr.17: 29562965-29562966 on Build GRCh38
rs777197003 Chr.17: 29563058-29563058 on Build GRCh38
rs374130392 Chr.17: 29562952-29562952 on Build GRCh38
rs745717342 Chr.17: 29563092-29563092 on Build GRCh38
rs780968261 Chr.17: 29563082-29563082 on Build GRCh38
rs760626515 Chr.17: 29563031-29563032 on Build GRCh38
rs200321208 Chr.17: 29562950-29562950 on Build GRCh38
rs368025165 Chr.17: 29563094-29563094 on Build GRCh38
rs1024270072 Chr.17: 29563002-29563002 on Build GRCh38
rs145924627 Chr.17: 29563064-29563064 on Build GRCh38
rs911742675 Chr.17: 29562993-29562993 on Build GRCh38
rs757913113 Chr.17: 29563054-29563054 on Build GRCh38
rs139415857 Chr.17: 29562980-29562980 on Build GRCh38
rs374284598 Chr.17: 29562920-29562920 on Build GRCh38
rs773846572 Chr.17: 29562918-29562918 on Build GRCh38

View Full Product Details