Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108605992-108606253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102B
Entrez Gene ID
284611
Gene Name
family with sequence similarity 102 member B
Gene Aliases
SYM-3B
UniGene
Hs.200230
Gene Chromosome Location
Chr.1: 108560349-108639327 on Build GRCh38
Transcripts
NM_001010883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936202 c.176G>C p.G59A Chr.1: 108606233-108606233 on Build GRCh38
4936201 c.269G>C p.G90A Chr.1: 108606233-108606233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573175058 Chr.1: 108606215-108606216 on Build GRCh38
rs889772032 Chr.1: 108606177-108606177 on Build GRCh38
rs762864380 Chr.1: 108606226-108606226 on Build GRCh38
rs763971676 Chr.1: 108606095-108606095 on Build GRCh38
rs1003631792 Chr.1: 108606164-108606164 on Build GRCh38
rs765441763 Chr.1: 108606128-108606128 on Build GRCh38
rs776294260 Chr.1: 108606187-108606187 on Build GRCh38
rs879051462 Chr.1: 108606126-108606126 on Build GRCh38
rs750515837 Chr.1: 108606165-108606165 on Build GRCh38
rs774997649 Chr.1: 108606213-108606213 on Build GRCh38
rs1053534131 Chr.1: 108606100-108606100 on Build GRCh38
rs769302598 Chr.1: 108606188-108606188 on Build GRCh38
rs571713077 Chr.1: 108606097-108606097 on Build GRCh38
rs374813921 Chr.1: 108606183-108606183 on Build GRCh38
rs77557583 Chr.1: 108606063-108606063 on Build GRCh38
rs906163610 Chr.1: 108606133-108606133 on Build GRCh38
rs780782018 Chr.1: 108606099-108606099 on Build GRCh38
rs983733283 Chr.1: 108606179-108606179 on Build GRCh38
rs534961308 Chr.1: 108606052-108606052 on Build GRCh38
rs79000760 Chr.1: 108606103-108606103 on Build GRCh38
rs933320213 Chr.1: 108606064-108606064 on Build GRCh38
rs371227136 Chr.1: 108606172-108606172 on Build GRCh38
rs762060573 Chr.1: 108606125-108606125 on Build GRCh38

View Full Product Details