Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40224605-40224848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTD2
Entrez Gene ID
79935
Gene Name
cyclin N-terminal domain containing 2
Gene Aliases
CCNP
UniGene
Hs.631603
Gene Chromosome Location
Chr.19: 40222208-40227111 on Build GRCh38
Transcripts
NM_024877.3 XM_006723395.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5782061 c.275G>A p.R92H Chr.19: 40224804-40224804 on Build GRCh38
6004210 c.271T>A p.Y91N Chr.19: 40224640-40224640 on Build GRCh38
6465774 c.357+5G>A p.? Chr.19: 40224717-40224717 on Build GRCh38
6151077 c.280C>G p.L94V Chr.19: 40224631-40224631 on Build GRCh38
3970987 c.347T>G p.V116G Chr.19: 40224732-40224732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749146083 Chr.19: 40224726-40224726 on Build GRCh38
rs780681284 Chr.19: 40224714-40224714 on Build GRCh38
rs182797546 Chr.19: 40224673-40224673 on Build GRCh38
rs759579240 Chr.19: 40224739-40224739 on Build GRCh38
rs1047060501 Chr.19: 40224815-40224815 on Build GRCh38
rs368367431 Chr.19: 40224659-40224659 on Build GRCh38
rs770967687 Chr.19: 40224666-40224666 on Build GRCh38
rs779518174 Chr.19: 40224704-40224704 on Build GRCh38
rs752269898 Chr.19: 40224820-40224820 on Build GRCh38
rs777426574 Chr.19: 40224719-40224719 on Build GRCh38
rs764312304 Chr.19: 40224687-40224687 on Build GRCh38
rs760969486 Chr.19: 40224765-40224765 on Build GRCh38
rs780663089 Chr.19: 40224805-40224805 on Build GRCh38
rs930016969 Chr.19: 40224797-40224797 on Build GRCh38
rs774263091 Chr.19: 40224667-40224667 on Build GRCh38
rs760984116 Chr.19: 40224686-40224686 on Build GRCh38
rs143673271 Chr.19: 40224651-40224651 on Build GRCh38
rs148388441 Chr.19: 40224627-40224627 on Build GRCh38
rs775726631 Chr.19: 40224754-40224754 on Build GRCh38
rs920034589 Chr.19: 40224777-40224777 on Build GRCh38
rs777835270 Chr.19: 40224764-40224764 on Build GRCh38
rs563012956 Chr.19: 40224676-40224676 on Build GRCh38
rs530544986 Chr.19: 40224690-40224690 on Build GRCh38
rs745908551 Chr.19: 40224776-40224776 on Build GRCh38
rs955686544 Chr.19: 40224680-40224680 on Build GRCh38
rs987041295 Chr.19: 40224693-40224693 on Build GRCh38
rs370943753 Chr.19: 40224696-40224696 on Build GRCh38
rs1001276678 Chr.19: 40224710-40224710 on Build GRCh38
rs747736892 Chr.19: 40224716-40224716 on Build GRCh38
rs987452772 Chr.19: 40224793-40224793 on Build GRCh38
rs375099334 Chr.19: 40224653-40224653 on Build GRCh38
rs754123618 Chr.19: 40224769-40224769 on Build GRCh38
rs750859975 Chr.19: 40224802-40224802 on Build GRCh38
rs199816045 Chr.19: 40224732-40224732 on Build GRCh38
rs762786202 Chr.19: 40224660-40224660 on Build GRCh38
rs769492676 Chr.19: 40224718-40224718 on Build GRCh38
rs759737417 Chr.19: 40224668-40224668 on Build GRCh38
rs754441720 Chr.19: 40224709-40224709 on Build GRCh38
rs770750605 Chr.19: 40224729-40224729 on Build GRCh38
rs765548200 Chr.19: 40224785-40224785 on Build GRCh38
rs552280466 Chr.19: 40224744-40224744 on Build GRCh38
rs750913915 Chr.19: 40224708-40224708 on Build GRCh38
rs1026195807 Chr.19: 40224767-40224767 on Build GRCh38

View Full Product Details