Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18596557-18596782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF1
Entrez Gene ID
9244
Gene Name
cytokine receptor like factor 1
Gene Aliases
CISS, CISS1, CLF, CLF-1, NR6, zcytor5
UniGene
Hs.114948
Gene Chromosome Location
Chr.19: 18593225-18606850 on Build GRCh38
Transcripts
NM_004750.4 XM_011528422.2 XM_011528423.2 XM_011528424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993125 c.973G>A p.G325R Chr.19: 18596673-18596673 on Build GRCh38
4674735 c.1001C>G p.T334R Chr.19: 18596645-18596645 on Build GRCh38
4953011 c.917T>C p.V306A Chr.19: 18596729-18596729 on Build GRCh38
5484259 c.897C>T p.A299A Chr.19: 18596749-18596749 on Build GRCh38
4075705 c.888C>A p.C296* Chr.19: 18596758-18596758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779981519 Chr.19: 18596706-18596706 on Build GRCh38
rs772871567 Chr.19: 18596635-18596635 on Build GRCh38
rs1010700288 Chr.19: 18596641-18596641 on Build GRCh38
rs758698603 Chr.19: 18596674-18596674 on Build GRCh38
rs747706339 Chr.19: 18596734-18596734 on Build GRCh38
rs779014706 Chr.19: 18596588-18596588 on Build GRCh38
rs909562748 Chr.19: 18596632-18596632 on Build GRCh38
rs769748607 Chr.19: 18596595-18596595 on Build GRCh38
rs745787497 Chr.19: 18596590-18596590 on Build GRCh38
rs926518620 Chr.19: 18596627-18596627 on Build GRCh38
rs761565793 Chr.19: 18596616-18596616 on Build GRCh38
rs147530893 Chr.19: 18596589-18596589 on Build GRCh38
rs768299320 Chr.19: 18596611-18596611 on Build GRCh38
rs35459448 Chr.19: 18596626-18596626 on Build GRCh38
rs772777727 Chr.19: 18596742-18596742 on Build GRCh38
rs752834862 Chr.19: 18596670-18596670 on Build GRCh38
rs755980118 Chr.19: 18596686-18596686 on Build GRCh38
rs755157766 Chr.19: 18596587-18596587 on Build GRCh38
rs754193561 Chr.19: 18596659-18596659 on Build GRCh38
rs765500879 Chr.19: 18596757-18596757 on Build GRCh38
rs771881717 Chr.19: 18596736-18596736 on Build GRCh38
rs372647247 Chr.19: 18596612-18596612 on Build GRCh38
rs749087012 Chr.19: 18596610-18596610 on Build GRCh38
rs766799203 Chr.19: 18596581-18596581 on Build GRCh38
rs777271351 Chr.19: 18596756-18596756 on Build GRCh38
rs975326391 Chr.19: 18596715-18596715 on Build GRCh38
rs770620678 Chr.19: 18596746-18596746 on Build GRCh38
rs766994193 Chr.19: 18596647-18596647 on Build GRCh38
rs754221400 Chr.19: 18596613-18596613 on Build GRCh38
rs768303738 Chr.19: 18596712-18596712 on Build GRCh38
rs529563490 Chr.19: 18596660-18596660 on Build GRCh38
rs1022042842 Chr.19: 18596656-18596656 on Build GRCh38
rs149763685 Chr.19: 18596743-18596743 on Build GRCh38
rs562107248 Chr.19: 18596761-18596761 on Build GRCh38
rs10402368 Chr.19: 18596580-18596580 on Build GRCh38
rs532927701 Chr.19: 18596600-18596600 on Build GRCh38
rs983769321 Chr.19: 18596629-18596629 on Build GRCh38
rs879869560 Chr.19: 18596673-18596673 on Build GRCh38
rs777786780 Chr.19: 18596675-18596675 on Build GRCh38
rs1007785837 Chr.19: 18596721-18596721 on Build GRCh38
rs771459625 Chr.19: 18596628-18596628 on Build GRCh38
rs922416641 Chr.19: 18596664-18596664 on Build GRCh38
rs546500486 Chr.19: 18596682-18596682 on Build GRCh38
rs777524208 Chr.19: 18596616-18596617 on Build GRCh38
rs778815231 Chr.19: 18596730-18596730 on Build GRCh38
rs765697276 Chr.19: 18596669-18596669 on Build GRCh38
rs756815834 Chr.19: 18596657-18596660 on Build GRCh38
rs746478763 Chr.19: 18596621-18596621 on Build GRCh38
rs376384592 Chr.19: 18596640-18596640 on Build GRCh38
rs137853933 Chr.19: 18596711-18596711 on Build GRCh38
rs1003608922 Chr.19: 18596605-18596605 on Build GRCh38

View Full Product Details