Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151253122-151253383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GM2A
Entrez Gene ID
2760
Gene Name
GM2 ganglioside activator
Gene Aliases
GM2-AP, SAP-3
UniGene
Hs.483873
Gene Chromosome Location
Chr.5: 151253052-151270394 on Build GRCh38
Transcripts
NM_000405.4 NM_001167607.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3244691 c.18G>A p.Q6Q Chr.5: 151253234-151253234 on Build GRCh38
6169955 c.25C>T p.L9F Chr.5: 151253241-151253241 on Build GRCh38
3761040 c.55G>A p.A19T Chr.5: 151253271-151253271 on Build GRCh38
4481349 c.24C>T p.P8P Chr.5: 151253240-151253240 on Build GRCh38
462034 c.51C>T p.L17L Chr.5: 151253267-151253267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368443364 Chr.5: 151253301-151253301 on Build GRCh38
rs751233771 Chr.5: 151253175-151253175 on Build GRCh38
rs1046381987 Chr.5: 151253357-151253357 on Build GRCh38
rs557461469 Chr.5: 151253229-151253229 on Build GRCh38
rs781089525 Chr.5: 151253323-151253326 on Build GRCh38
rs185316713 Chr.5: 151253324-151253324 on Build GRCh38
rs763602251 Chr.5: 151253194-151253195 on Build GRCh38
rs772342276 Chr.5: 151253218-151253218 on Build GRCh38
rs975000914 Chr.5: 151253325-151253325 on Build GRCh38
rs753408544 Chr.5: 151253343-151253343 on Build GRCh38
rs143867953 Chr.5: 151253249-151253249 on Build GRCh38
rs1048718 Chr.5: 151253176-151253176 on Build GRCh38
rs543252624 Chr.5: 151253250-151253250 on Build GRCh38
rs28365994 Chr.5: 151253294-151253294 on Build GRCh38
rs907141181 Chr.5: 151253359-151253359 on Build GRCh38
rs749782834 Chr.5: 151253199-151253199 on Build GRCh38
rs1048719 Chr.5: 151253271-151253271 on Build GRCh38
rs768756904 Chr.5: 151253285-151253285 on Build GRCh38
rs762676667 Chr.5: 151253227-151253227 on Build GRCh38
rs767131690 Chr.5: 151253232-151253232 on Build GRCh38
rs936770728 Chr.5: 151253278-151253278 on Build GRCh38
rs372781289 Chr.5: 151253331-151253331 on Build GRCh38
rs909177693 Chr.5: 151253296-151253296 on Build GRCh38
rs764022141 Chr.5: 151253231-151253231 on Build GRCh38
rs529126687 Chr.5: 151253279-151253279 on Build GRCh38
rs201933430 Chr.5: 151253323-151253323 on Build GRCh38
rs144932594 Chr.5: 151253173-151253173 on Build GRCh38
rs893045422 Chr.5: 151253213-151253213 on Build GRCh38
rs751251529 Chr.5: 151253207-151253208 on Build GRCh38
rs553449209 Chr.5: 151253186-151253186 on Build GRCh38
rs759364593 Chr.5: 151253318-151253318 on Build GRCh38
rs950802002 Chr.5: 151253293-151253293 on Build GRCh38
rs775585062 Chr.5: 151253220-151253220 on Build GRCh38
rs545467021 Chr.5: 151253224-151253224 on Build GRCh38
rs766243967 Chr.5: 151253228-151253228 on Build GRCh38
rs998937927 Chr.5: 151253164-151253164 on Build GRCh38
rs780278367 Chr.5: 151253273-151253273 on Build GRCh38
rs756968170 Chr.5: 151253208-151253208 on Build GRCh38
rs562249526 Chr.5: 151253221-151253221 on Build GRCh38
rs200545376 Chr.5: 151253272-151253272 on Build GRCh38
rs752323907 Chr.5: 151253240-151253240 on Build GRCh38
rs755584488 Chr.5: 151253195-151253195 on Build GRCh38
rs371628812 Chr.5: 151253210-151253210 on Build GRCh38
rs111913684 Chr.5: 151253298-151253298 on Build GRCh38
rs779239922 Chr.5: 151253261-151253261 on Build GRCh38
rs1026835435 Chr.5: 151253275-151253275 on Build GRCh38
rs374161170 Chr.5: 151253255-151253255 on Build GRCh38
rs759249002 Chr.5: 151253230-151253230 on Build GRCh38
rs367935217 Chr.5: 151253179-151253179 on Build GRCh38
rs760185979 Chr.5: 151253243-151253243 on Build GRCh38
rs763659715 Chr.5: 151253336-151253336 on Build GRCh38
rs572025687 Chr.5: 151253215-151253215 on Build GRCh38

View Full Product Details