Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37011119-37011352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47C
Entrez Gene ID
442444
Gene Name
family with sequence similarity 47 member C
Gene Aliases
-
UniGene
Hs.535216
Gene Chromosome Location
Chr.X: 37008359-37011666 on Build GRCh38
Transcripts
NM_001013736.2

SNP Detail

SNP ID Chromosome Location
rs782432221 Chr.X: 37011245-37011245 on Build GRCh38
rs782657203 Chr.X: 37011242-37011242 on Build GRCh38
rs375998369 Chr.X: 37011273-37011273 on Build GRCh38
rs782041320 Chr.X: 37011331-37011331 on Build GRCh38
rs782569583 Chr.X: 37011200-37011200 on Build GRCh38
rs781894377 Chr.X: 37011189-37011189 on Build GRCh38
rs1995914 Chr.X: 37011181-37011181 on Build GRCh38
rs782191586 Chr.X: 37011250-37011250 on Build GRCh38
rs201493707 Chr.X: 37011280-37011280 on Build GRCh38
rs782306470 Chr.X: 37011252-37011252 on Build GRCh38
rs782240419 Chr.X: 37011243-37011243 on Build GRCh38
rs782457615 Chr.X: 37011197-37011197 on Build GRCh38
rs782046931 Chr.X: 37011165-37011165 on Build GRCh38
rs41311787 Chr.X: 37011320-37011320 on Build GRCh38
rs61730906 Chr.X: 37011183-37011183 on Build GRCh38
rs782790091 Chr.X: 37011163-37011163 on Build GRCh38
rs782681825 Chr.X: 37011186-37011186 on Build GRCh38
rs373029478 Chr.X: 37011239-37011239 on Build GRCh38
rs782008490 Chr.X: 37011249-37011249 on Build GRCh38
rs139172934 Chr.X: 37011160-37011160 on Build GRCh38
rs781799672 Chr.X: 37011164-37011164 on Build GRCh38
rs782417722 Chr.X: 37011317-37011318 on Build GRCh38
rs782235862 Chr.X: 37011212-37011212 on Build GRCh38

View Full Product Details