Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88805503-88805728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDT1
Entrez Gene ID
81620
Gene Name
chromatin licensing and DNA replication factor 1
Gene Aliases
DUP, RIS2
UniGene
Hs.122908
Gene Chromosome Location
Chr.16: 88803778-88809258 on Build GRCh38
Transcripts
NM_030928.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063521 c.638C>T p.T213M Chr.16: 88805589-88805589 on Build GRCh38
5754946 c.562T>C p.Y188H Chr.16: 88805513-88805513 on Build GRCh38
974766 c.634G>A p.E212K Chr.16: 88805585-88805585 on Build GRCh38
7284459 c.667G>A p.V223I Chr.16: 88805618-88805618 on Build GRCh38
6625822 c.552C>T p.L184L Chr.16: 88805503-88805503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145552478 Chr.16: 88805564-88805564 on Build GRCh38
rs572456795 Chr.16: 88805688-88805688 on Build GRCh38
rs376711413 Chr.16: 88805590-88805590 on Build GRCh38
rs548341429 Chr.16: 88805584-88805584 on Build GRCh38
rs750025399 Chr.16: 88805634-88805634 on Build GRCh38
rs749876459 Chr.16: 88805550-88805550 on Build GRCh38
rs140700251 Chr.16: 88805543-88805543 on Build GRCh38
rs377591661 Chr.16: 88805706-88805706 on Build GRCh38
rs956776049 Chr.16: 88805687-88805688 on Build GRCh38
rs372330019 Chr.16: 88805670-88805670 on Build GRCh38
rs767922745 Chr.16: 88805560-88805560 on Build GRCh38
rs538165836 Chr.16: 88805633-88805633 on Build GRCh38
rs570089762 Chr.16: 88805621-88805621 on Build GRCh38
rs767280942 Chr.16: 88805699-88805699 on Build GRCh38
rs997921410 Chr.16: 88805616-88805631 on Build GRCh38
rs762869154 Chr.16: 88805611-88805611 on Build GRCh38
rs777434586 Chr.16: 88805538-88805538 on Build GRCh38
rs373340835 Chr.16: 88805618-88805618 on Build GRCh38
rs150960744 Chr.16: 88805533-88805533 on Build GRCh38
rs201758940 Chr.16: 88805708-88805708 on Build GRCh38
rs750158020 Chr.16: 88805705-88805705 on Build GRCh38
rs753375373 Chr.16: 88805710-88805710 on Build GRCh38
rs980887091 Chr.16: 88805579-88805579 on Build GRCh38
rs769385427 Chr.16: 88805585-88805585 on Build GRCh38
rs535723934 Chr.16: 88805665-88805665 on Build GRCh38
rs753832416 Chr.16: 88805565-88805565 on Build GRCh38
rs756222380 Chr.16: 88805528-88805528 on Build GRCh38
rs764018742 Chr.16: 88805612-88805612 on Build GRCh38
rs746736930 Chr.16: 88805582-88805582 on Build GRCh38
rs752647897 Chr.16: 88805601-88805601 on Build GRCh38
rs750567415 Chr.16: 88805525-88805525 on Build GRCh38
rs146872515 Chr.16: 88805589-88805589 on Build GRCh38
rs544489602 Chr.16: 88805532-88805532 on Build GRCh38
rs780557868 Chr.16: 88805636-88805637 on Build GRCh38
rs752592573 Chr.16: 88805569-88805569 on Build GRCh38
rs75439984 Chr.16: 88805574-88805574 on Build GRCh38
rs774947382 Chr.16: 88805557-88805557 on Build GRCh38
rs201871319 Chr.16: 88805594-88805594 on Build GRCh38
rs758197391 Chr.16: 88805570-88805570 on Build GRCh38
rs769924210 Chr.16: 88805681-88805681 on Build GRCh38
rs777377390 Chr.16: 88805572-88805572 on Build GRCh38
rs761513270 Chr.16: 88805698-88805698 on Build GRCh38
rs542997068 Chr.16: 88805613-88805613 on Build GRCh38
rs765182594 Chr.16: 88805568-88805568 on Build GRCh38
rs377465796 Chr.16: 88805630-88805630 on Build GRCh38
rs201106735 Chr.16: 88805647-88805647 on Build GRCh38
rs773495319 Chr.16: 88805561-88805561 on Build GRCh38
rs747357665 Chr.16: 88805660-88805660 on Build GRCh38
rs775545437 Chr.16: 88805683-88805683 on Build GRCh38
rs370127987 Chr.16: 88805591-88805591 on Build GRCh38
rs368138332 Chr.16: 88805652-88805652 on Build GRCh38
rs374891839 Chr.16: 88805648-88805648 on Build GRCh38
rs755694518 Chr.16: 88805647-88805648 on Build GRCh38
rs754960043 Chr.16: 88805567-88805567 on Build GRCh38
rs189449467 Chr.16: 88805646-88805646 on Build GRCh38
rs747918319 Chr.16: 88805534-88805534 on Build GRCh38
rs760793444 Chr.16: 88805563-88805563 on Build GRCh38
rs758141929 Chr.16: 88805537-88805537 on Build GRCh38
rs765235636 Chr.16: 88805600-88805600 on Build GRCh38
rs755695035 Chr.16: 88805642-88805642 on Build GRCh38
rs1000923703 Chr.16: 88805664-88805664 on Build GRCh38
rs199632777 Chr.16: 88805694-88805694 on Build GRCh38
rs377399257 Chr.16: 88805687-88805687 on Build GRCh38
rs775994089 Chr.16: 88805544-88805544 on Build GRCh38
rs760995727 Chr.16: 88805593-88805593 on Build GRCh38
rs551760059 Chr.16: 88805596-88805596 on Build GRCh38
rs1016226625 Chr.16: 88805608-88805608 on Build GRCh38
rs755010649 Chr.16: 88805606-88805617 on Build GRCh38
rs766683758 Chr.16: 88805542-88805542 on Build GRCh38
rs769261147 Chr.16: 88805556-88805556 on Build GRCh38
rs762294762 Chr.16: 88805559-88805559 on Build GRCh38
rs200871594 Chr.16: 88805679-88805679 on Build GRCh38
rs752304125 Chr.16: 88805648-88805675 on Build GRCh38
rs373709218 Chr.16: 88805530-88805530 on Build GRCh38
rs934297539 Chr.16: 88805704-88805704 on Build GRCh38

View Full Product Details