Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44287286-44287559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRE
Entrez Gene ID
326
Gene Name
autoimmune regulator
Gene Aliases
AIRE1, APECED, APS1, APSI, PGA1
UniGene
Hs.129829
Gene Chromosome Location
Chr.21: 44285838-44298219 on Build GRCh38
Transcripts
NM_000383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415442 c.490A>G p.K164E Chr.21: 44287543-44287543 on Build GRCh38
7322417 c.497C>T p.P166L Chr.21: 44287550-44287550 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs56363875 Chr.21: 44287433-44287433 on Build GRCh38
rs771592755 Chr.21: 44287541-44287541 on Build GRCh38
rs958295959 Chr.21: 44287434-44287434 on Build GRCh38
rs551213280 Chr.21: 44287483-44287483 on Build GRCh38
rs978361635 Chr.21: 44287311-44287311 on Build GRCh38
rs989518616 Chr.21: 44287435-44287435 on Build GRCh38
rs768310786 Chr.21: 44287510-44287510 on Build GRCh38
rs936649259 Chr.21: 44287335-44287335 on Build GRCh38
rs1032171597 Chr.21: 44287515-44287515 on Build GRCh38
rs560928264 Chr.21: 44287413-44287413 on Build GRCh38
rs56171459 Chr.21: 44287446-44287446 on Build GRCh38
rs967513511 Chr.21: 44287406-44287406 on Build GRCh38
rs767639114 Chr.21: 44287319-44287319 on Build GRCh38
rs1006583932 Chr.21: 44287419-44287419 on Build GRCh38
rs965518051 Chr.21: 44287423-44287423 on Build GRCh38
rs1051919739 Chr.21: 44287447-44287447 on Build GRCh38
rs779080108 Chr.21: 44287359-44287359 on Build GRCh38
rs867979277 Chr.21: 44287537-44287537 on Build GRCh38
rs773945988 Chr.21: 44287511-44287511 on Build GRCh38
rs942278021 Chr.21: 44287436-44287436 on Build GRCh38
rs112759342 Chr.21: 44287412-44287412 on Build GRCh38
rs540855212 Chr.21: 44287424-44287424 on Build GRCh38
rs746101086 Chr.21: 44287536-44287537 on Build GRCh38
rs528176573 Chr.21: 44287415-44287415 on Build GRCh38
rs575140436 Chr.21: 44287396-44287396 on Build GRCh38
rs138949170 Chr.21: 44287538-44287538 on Build GRCh38
rs957939181 Chr.21: 44287395-44287395 on Build GRCh38
rs888920898 Chr.21: 44287345-44287345 on Build GRCh38
rs1007486307 Chr.21: 44287357-44287357 on Build GRCh38
rs146020455 Chr.21: 44287404-44287404 on Build GRCh38
rs117917332 Chr.21: 44287334-44287334 on Build GRCh38
rs1036910945 Chr.21: 44287398-44287398 on Build GRCh38
rs897605743 Chr.21: 44287307-44287307 on Build GRCh38
rs938495466 Chr.21: 44287471-44287471 on Build GRCh38
rs764644932 Chr.21: 44287441-44287441 on Build GRCh38
rs532734160 Chr.21: 44287444-44287444 on Build GRCh38
rs997660134 Chr.21: 44287308-44287308 on Build GRCh38
rs1019776667 Chr.21: 44287422-44287422 on Build GRCh38
rs888959872 Chr.21: 44287461-44287461 on Build GRCh38
rs1054291148 Chr.21: 44287342-44287342 on Build GRCh38
rs746399817 Chr.21: 44287380-44287380 on Build GRCh38
rs569160989 Chr.21: 44287503-44287503 on Build GRCh38
rs1007440250 Chr.21: 44287481-44287481 on Build GRCh38
rs916642248 Chr.21: 44287353-44287353 on Build GRCh38
rs781470144 Chr.21: 44287532-44287537 on Build GRCh38
rs937472076 Chr.21: 44287416-44287416 on Build GRCh38

View Full Product Details