Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8009407-8009678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY2D
Entrez Gene ID
3000
Gene Name
guanylate cyclase 2D, retinal
Gene Aliases
CORD5, CORD6, CYGD, GUC1A4, GUC2D, LCA, LCA1, RCD2, RETGC-1, ROS-GC1, ROSGC, retGC
UniGene
Hs.592109
Gene Chromosome Location
Chr.17: 8002670-8020340 on Build GRCh38
Transcripts
NM_000180.3 XM_011523816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437763 c.1719C>T p.I573I Chr.17: 8009556-8009556 on Build GRCh38
1253916 c.1721G>A p.R574H Chr.17: 8009558-8009558 on Build GRCh38
135736 c.1669G>A p.G557R Chr.17: 8009506-8009506 on Build GRCh38
7191231 c.1729A>G p.T577A Chr.17: 8009566-8009566 on Build GRCh38
4139938 c.1674C>T p.D558D Chr.17: 8009511-8009511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774554121 Chr.17: 8009559-8009559 on Build GRCh38
rs761529451 Chr.17: 8009492-8009492 on Build GRCh38
rs761631031 Chr.17: 8009574-8009574 on Build GRCh38
rs933391884 Chr.17: 8009645-8009645 on Build GRCh38
rs371322812 Chr.17: 8009456-8009456 on Build GRCh38
rs1035738652 Chr.17: 8009444-8009444 on Build GRCh38
rs751811525 Chr.17: 8009499-8009499 on Build GRCh38
rs745533728 Chr.17: 8009553-8009553 on Build GRCh38
rs372118691 Chr.17: 8009457-8009457 on Build GRCh38
rs760662744 Chr.17: 8009476-8009476 on Build GRCh38
rs766557426 Chr.17: 8009493-8009493 on Build GRCh38
rs61749755 Chr.17: 8009531-8009531 on Build GRCh38
rs886043672 Chr.17: 8009550-8009550 on Build GRCh38
rs951042742 Chr.17: 8009548-8009548 on Build GRCh38
rs28743021 Chr.17: 8009561-8009561 on Build GRCh38
rs563093147 Chr.17: 8009445-8009445 on Build GRCh38
rs760681687 Chr.17: 8009625-8009625 on Build GRCh38
rs767561967 Chr.17: 8009503-8009503 on Build GRCh38
rs759723474 Chr.17: 8009597-8009597 on Build GRCh38
rs968943154 Chr.17: 8009485-8009485 on Build GRCh38
rs568220391 Chr.17: 8009544-8009544 on Build GRCh38
rs546875049 Chr.17: 8009537-8009537 on Build GRCh38
rs775192787 Chr.17: 8009471-8009471 on Build GRCh38
rs757259115 Chr.17: 8009496-8009497 on Build GRCh38
rs933462730 Chr.17: 8009545-8009545 on Build GRCh38
rs560270873 Chr.17: 8009558-8009558 on Build GRCh38
rs764591974 Chr.17: 8009626-8009626 on Build GRCh38
rs188568530 Chr.17: 8009509-8009509 on Build GRCh38
rs1013955758 Chr.17: 8009478-8009478 on Build GRCh38
rs752758326 Chr.17: 8009624-8009624 on Build GRCh38
rs371458632 Chr.17: 8009636-8009636 on Build GRCh38
rs756639026 Chr.17: 8009536-8009536 on Build GRCh38
rs754170292 Chr.17: 8009539-8009539 on Build GRCh38
rs377287956 Chr.17: 8009556-8009556 on Build GRCh38
rs61749756 Chr.17: 8009554-8009554 on Build GRCh38
rs759564156 Chr.17: 8009502-8009502 on Build GRCh38
rs778664525 Chr.17: 8009551-8009551 on Build GRCh38
rs570329813 Chr.17: 8009489-8009489 on Build GRCh38
rs745306850 Chr.17: 8009459-8009459 on Build GRCh38
rs374800328 Chr.17: 8009458-8009458 on Build GRCh38
rs771734877 Chr.17: 8009467-8009467 on Build GRCh38
rs137853897 Chr.17: 8009557-8009557 on Build GRCh38
rs376062280 Chr.17: 8009612-8009612 on Build GRCh38
rs754329901 Chr.17: 8009628-8009628 on Build GRCh38
rs757589496 Chr.17: 8009631-8009631 on Build GRCh38
rs776439113 Chr.17: 8009568-8009568 on Build GRCh38
rs180896227 Chr.17: 8009635-8009635 on Build GRCh38
rs995374873 Chr.17: 8009646-8009646 on Build GRCh38
rs557073963 Chr.17: 8009589-8009589 on Build GRCh38

View Full Product Details