Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37129794-37130033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3437480 c.106C>T p.L36L Chr.10: 37129945-37129945 on Build GRCh38
4521015 c.108G>A p.L36L Chr.10: 37129947-37129947 on Build GRCh38
1956208 c.77T>C p.V26A Chr.10: 37129916-37129916 on Build GRCh38
684413 c.61C>T p.L21L Chr.10: 37129900-37129900 on Build GRCh38
5800824 c.144C>G p.G48G Chr.10: 37129983-37129983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746029797 Chr.10: 37129883-37129883 on Build GRCh38
rs750282260 Chr.10: 37129967-37129967 on Build GRCh38
rs565676057 Chr.10: 37129933-37129933 on Build GRCh38
rs989567967 Chr.10: 37129828-37129828 on Build GRCh38
rs534812137 Chr.10: 37129936-37129936 on Build GRCh38
rs749051263 Chr.10: 37129858-37129858 on Build GRCh38
rs369569719 Chr.10: 37129983-37129983 on Build GRCh38
rs775423794 Chr.10: 37129888-37129888 on Build GRCh38
rs200651327 Chr.10: 37129984-37129984 on Build GRCh38
rs376005400 Chr.10: 37130001-37130001 on Build GRCh38
rs778699750 Chr.10: 37129882-37129882 on Build GRCh38
rs747032437 Chr.10: 37129982-37129982 on Build GRCh38
rs373549644 Chr.10: 37129889-37129889 on Build GRCh38
rs760271901 Chr.10: 37129940-37129940 on Build GRCh38
rs778609658 Chr.10: 37129958-37129958 on Build GRCh38
rs532175584 Chr.10: 37129894-37129894 on Build GRCh38
rs752257064 Chr.10: 37129852-37129852 on Build GRCh38
rs377486764 Chr.10: 37129975-37129975 on Build GRCh38
rs899935311 Chr.10: 37129998-37129998 on Build GRCh38
rs748609132 Chr.10: 37129987-37129987 on Build GRCh38
rs779403347 Chr.10: 37129887-37129888 on Build GRCh38
rs770713635 Chr.10: 37129971-37129971 on Build GRCh38
rs777814728 Chr.10: 37129856-37129856 on Build GRCh38
rs753724879 Chr.10: 37129949-37129949 on Build GRCh38
rs947655450 Chr.10: 37129906-37129906 on Build GRCh38
rs914882787 Chr.10: 37129867-37129867 on Build GRCh38
rs777322288 Chr.10: 37129892-37129892 on Build GRCh38
rs568301150 Chr.10: 37129989-37129989 on Build GRCh38
rs757005423 Chr.10: 37129861-37129861 on Build GRCh38
rs763623721 Chr.10: 37129948-37129948 on Build GRCh38
rs754702484 Chr.10: 37129844-37129844 on Build GRCh38
rs770236719 Chr.10: 37129898-37129898 on Build GRCh38
rs772339444 Chr.10: 37129886-37129886 on Build GRCh38
rs1041973101 Chr.10: 37129908-37129908 on Build GRCh38
rs756001489 Chr.10: 37129854-37129854 on Build GRCh38
rs759045913 Chr.10: 37129918-37129918 on Build GRCh38
rs552288556 Chr.10: 37129911-37129911 on Build GRCh38
rs766880945 Chr.10: 37129848-37129848 on Build GRCh38
rs763100757 Chr.10: 37129952-37129952 on Build GRCh38

View Full Product Details