Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231206150-231206401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC9
Entrez Gene ID
80210
Gene Name
armadillo repeat containing 9
Gene Aliases
ARM, KU-MEL-1, NS21
UniGene
Hs.162411
Gene Chromosome Location
Chr.2: 231198546-231394991 on Build GRCh38
Transcripts
NM_001271466.2 NM_001291656.1 NM_025139.5 XM_011511905.1 XM_011511906.1 XM_011511907.1 XM_011511908.2 XM_011511909.2 XM_011511910.1 XM_011511911.1 XM_011511912.1 XM_011511913.2 XM_011511914.2 XM_011511915.2 XM_011511916.2 XM_017005018.1 XM_017005019.1 XM_017005020.1 XM_017005021.1 XM_017005022.1 XM_017005023.1 XM_017005024.1 XM_017005025.1 XM_017005026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5750129 c.8A>G p.D3G Chr.2: 231206246-231206246 on Build GRCh38
5732333 c.51+1G>C p.? Chr.2: 231206290-231206290 on Build GRCh38
3838828 c.1A>G p.M1V Chr.2: 231206239-231206239 on Build GRCh38
1406167 c.3G>C p.M1I Chr.2: 231206241-231206241 on Build GRCh38
4801905 c.3G>T p.M1I Chr.2: 231206241-231206241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771993098 Chr.2: 231206336-231206337 on Build GRCh38
rs74651652 Chr.2: 231206193-231206193 on Build GRCh38
rs539065720 Chr.2: 231206179-231206179 on Build GRCh38
rs373735875 Chr.2: 231206309-231206309 on Build GRCh38
rs760205337 Chr.2: 231206304-231206304 on Build GRCh38
rs748497564 Chr.2: 231206266-231206266 on Build GRCh38
rs763945498 Chr.2: 231206339-231206339 on Build GRCh38
rs535278701 Chr.2: 231206181-231206181 on Build GRCh38
rs754818794 Chr.2: 231206231-231206231 on Build GRCh38
rs760386487 Chr.2: 231206211-231206211 on Build GRCh38
rs765740451 Chr.2: 231206212-231206212 on Build GRCh38
rs777503615 Chr.2: 231206240-231206241 on Build GRCh38
rs777727215 Chr.2: 231206239-231206239 on Build GRCh38
rs376958829 Chr.2: 231206251-231206251 on Build GRCh38
rs772906167 Chr.2: 231206209-231206209 on Build GRCh38
rs747197262 Chr.2: 231206337-231206338 on Build GRCh38
rs61753914 Chr.2: 231206248-231206248 on Build GRCh38
rs373513496 Chr.2: 231206236-231206236 on Build GRCh38
rs371428399 Chr.2: 231206201-231206201 on Build GRCh38
rs765258017 Chr.2: 231206319-231206319 on Build GRCh38
rs200925709 Chr.2: 231206257-231206257 on Build GRCh38
rs751396454 Chr.2: 231206340-231206340 on Build GRCh38
rs767650589 Chr.2: 231206273-231206273 on Build GRCh38
rs766074419 Chr.2: 231206306-231206306 on Build GRCh38
rs762102028 Chr.2: 231206287-231206287 on Build GRCh38
rs555508528 Chr.2: 231206344-231206344 on Build GRCh38
rs759123410 Chr.2: 231206316-231206316 on Build GRCh38
rs372983316 Chr.2: 231206328-231206328 on Build GRCh38
rs758332574 Chr.2: 231206237-231206237 on Build GRCh38
rs565988774 Chr.2: 231206204-231206204 on Build GRCh38
rs774906944 Chr.2: 231206263-231206263 on Build GRCh38
rs978107338 Chr.2: 231206283-231206283 on Build GRCh38
rs774676115 Chr.2: 231206195-231206195 on Build GRCh38
rs749276125 Chr.2: 231206334-231206335 on Build GRCh38
rs773571141 Chr.2: 231206296-231206296 on Build GRCh38
rs753418173 Chr.2: 231206215-231206215 on Build GRCh38
rs765310794 Chr.2: 231206233-231206233 on Build GRCh38
rs981230066 Chr.2: 231206183-231206183 on Build GRCh38
rs755948146 Chr.2: 231206214-231206217 on Build GRCh38
rs998309850 Chr.2: 231206224-231206224 on Build GRCh38
rs562937451 Chr.2: 231206246-231206246 on Build GRCh38
rs146897446 Chr.2: 231206208-231206208 on Build GRCh38
rs752627766 Chr.2: 231206322-231206322 on Build GRCh38
rs558020519 Chr.2: 231206213-231206213 on Build GRCh38
rs746014643 Chr.2: 231206241-231206241 on Build GRCh38
rs772362877 Chr.2: 231206286-231206286 on Build GRCh38

View Full Product Details