Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32615761-32616022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC73
Entrez Gene ID
493860
Gene Name
coiled-coil domain containing 73
Gene Aliases
NY-SAR-79
UniGene
Hs.706808
Gene Chromosome Location
Chr.11: 32602080-32794658 on Build GRCh38
Transcripts
NM_001008391.3 XM_011520139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926388 c.1352T>A p.F451Y Chr.11: 32615963-32615963 on Build GRCh38
6233353 c.1333G>A p.E445K Chr.11: 32615982-32615982 on Build GRCh38
6396574 c.1319A>G p.Y440C Chr.11: 32615996-32615996 on Build GRCh38
4987338 c.1309G>A p.D437N Chr.11: 32616006-32616006 on Build GRCh38
3415810 c.1296G>T p.E432D Chr.11: 32616019-32616019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760633040 Chr.11: 32615868-32615868 on Build GRCh38
rs375600920 Chr.11: 32615958-32615958 on Build GRCh38
rs774376275 Chr.11: 32615860-32615860 on Build GRCh38
rs761591957 Chr.11: 32615914-32615914 on Build GRCh38
rs552728381 Chr.11: 32615906-32615906 on Build GRCh38
rs970104354 Chr.11: 32615830-32615830 on Build GRCh38
rs758669232 Chr.11: 32615973-32615974 on Build GRCh38
rs763999903 Chr.11: 32615898-32615898 on Build GRCh38
rs200810856 Chr.11: 32615922-32615922 on Build GRCh38
rs938803090 Chr.11: 32615837-32615837 on Build GRCh38
rs751091078 Chr.11: 32615979-32615979 on Build GRCh38
rs766487815 Chr.11: 32615853-32615853 on Build GRCh38
rs781762552 Chr.11: 32615983-32615983 on Build GRCh38
rs913409307 Chr.11: 32615897-32615897 on Build GRCh38
rs771098685 Chr.11: 32615980-32615982 on Build GRCh38
rs980535579 Chr.11: 32615832-32615832 on Build GRCh38
rs950785822 Chr.11: 32615846-32615846 on Build GRCh38
rs766505077 Chr.11: 32615820-32615820 on Build GRCh38
rs187312854 Chr.11: 32615834-32615834 on Build GRCh38
rs1053092431 Chr.11: 32615883-32615883 on Build GRCh38
rs370055016 Chr.11: 32615962-32615962 on Build GRCh38
rs753130843 Chr.11: 32615843-32615845 on Build GRCh38
rs201707304 Chr.11: 32615985-32615985 on Build GRCh38
rs747079826 Chr.11: 32615974-32615974 on Build GRCh38
rs767480192 Chr.11: 32615856-32615856 on Build GRCh38
rs900773621 Chr.11: 32615964-32615964 on Build GRCh38
rs755581480 Chr.11: 32615982-32615982 on Build GRCh38
rs753673970 Chr.11: 32615903-32615903 on Build GRCh38
rs750105589 Chr.11: 32615923-32615923 on Build GRCh38

View Full Product Details