Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67053903-67054166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG1
Entrez Gene ID
786
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Gene Aliases
CACNLG
UniGene
Hs.147989
Gene Chromosome Location
Chr.17: 67044536-67056797 on Build GRCh38
Transcripts
NM_000727.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4068863 c.266G>A p.G89D Chr.17: 67054032-67054032 on Build GRCh38
1385376 c.286G>A p.E96K Chr.17: 67054052-67054052 on Build GRCh38
2797296 c.286G>T p.E96* Chr.17: 67054052-67054052 on Build GRCh38
159703 c.265G>A p.G89S Chr.17: 67054031-67054031 on Build GRCh38
6304588 c.268G>A p.E90K Chr.17: 67054034-67054034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202020889 Chr.17: 67054020-67054020 on Build GRCh38
rs758554446 Chr.17: 67054017-67054017 on Build GRCh38
rs866091801 Chr.17: 67054027-67054027 on Build GRCh38
rs753774873 Chr.17: 67054108-67054108 on Build GRCh38
rs899158821 Chr.17: 67053996-67053998 on Build GRCh38
rs375808344 Chr.17: 67053945-67053945 on Build GRCh38
rs920610146 Chr.17: 67054061-67054061 on Build GRCh38
rs200930741 Chr.17: 67054107-67054107 on Build GRCh38
rs774814035 Chr.17: 67054073-67054073 on Build GRCh38
rs751712868 Chr.17: 67054030-67054030 on Build GRCh38
rs774891661 Chr.17: 67053956-67053956 on Build GRCh38
rs777180382 Chr.17: 67054081-67054081 on Build GRCh38
rs766560706 Chr.17: 67054099-67054099 on Build GRCh38
rs765363207 Chr.17: 67053998-67053998 on Build GRCh38
rs766982478 Chr.17: 67053955-67053955 on Build GRCh38
rs750785147 Chr.17: 67054096-67054096 on Build GRCh38
rs368641628 Chr.17: 67054085-67054085 on Build GRCh38
rs375191225 Chr.17: 67053974-67053974 on Build GRCh38
rs777900825 Chr.17: 67054033-67054033 on Build GRCh38
rs371843031 Chr.17: 67054031-67054031 on Build GRCh38
rs745490109 Chr.17: 67054041-67054041 on Build GRCh38
rs774956064 Chr.17: 67054051-67054051 on Build GRCh38
rs1039122504 Chr.17: 67053992-67053992 on Build GRCh38
rs778573499 Chr.17: 67054040-67054040 on Build GRCh38
rs764631658 Chr.17: 67053965-67053965 on Build GRCh38
rs760216442 Chr.17: 67054052-67054052 on Build GRCh38
rs761117725 Chr.17: 67053962-67053962 on Build GRCh38
rs36047721 Chr.17: 67054084-67054085 on Build GRCh38
rs376099391 Chr.17: 67054042-67054042 on Build GRCh38
rs113718207 Chr.17: 67054071-67054071 on Build GRCh38
rs770820692 Chr.17: 67054039-67054039 on Build GRCh38
rs151274817 Chr.17: 67053939-67053939 on Build GRCh38
rs113550460 Chr.17: 67053924-67053924 on Build GRCh38
rs111986322 Chr.17: 67054072-67054072 on Build GRCh38
rs772851074 Chr.17: 67054088-67054088 on Build GRCh38
rs760059878 Chr.17: 67054114-67054115 on Build GRCh38
rs750599405 Chr.17: 67054010-67054010 on Build GRCh38
rs968109408 Chr.17: 67054087-67054087 on Build GRCh38
rs757168869 Chr.17: 67054112-67054112 on Build GRCh38
rs538457383 Chr.17: 67054105-67054105 on Build GRCh38
rs778716870 Chr.17: 67054116-67054116 on Build GRCh38
rs113103602 Chr.17: 67053994-67053994 on Build GRCh38
rs140431401 Chr.17: 67053953-67053953 on Build GRCh38
rs911725815 Chr.17: 67053980-67053980 on Build GRCh38
rs930638987 Chr.17: 67054065-67054065 on Build GRCh38
rs369092327 Chr.17: 67054098-67054098 on Build GRCh38
rs571080227 Chr.17: 67054093-67054093 on Build GRCh38
rs774123413 Chr.17: 67053954-67053954 on Build GRCh38
rs749094535 Chr.17: 67054034-67054034 on Build GRCh38

View Full Product Details