Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 21002524-21002797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOB
Entrez Gene ID
338
Gene Name
apolipoprotein B
Gene Aliases
FLDB, LDLCQ4, apoB-100, apoB-48
UniGene
Hs.120759
Gene Chromosome Location
Chr.2: 21001429-21044073 on Build GRCh38
Transcripts
NM_000384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4524348 c.12674G>A p.G4225E Chr.2: 21002748-21002748 on Build GRCh38
7242220 c.12862C>G p.Q4288E Chr.2: 21002560-21002560 on Build GRCh38
3577118 c.12738C>T p.F4246F Chr.2: 21002684-21002684 on Build GRCh38
5396097 c.12667G>A p.E4223K Chr.2: 21002755-21002755 on Build GRCh38
1183249 c.12895C>A p.L4299I Chr.2: 21002527-21002527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752558963 Chr.2: 21002649-21002649 on Build GRCh38
rs750204106 Chr.2: 21002571-21002571 on Build GRCh38
rs377125320 Chr.2: 21002724-21002724 on Build GRCh38
rs764903010 Chr.2: 21002559-21002559 on Build GRCh38
rs772632789 Chr.2: 21002614-21002614 on Build GRCh38
rs777838124 Chr.2: 21002602-21002602 on Build GRCh38
rs972943191 Chr.2: 21002592-21002592 on Build GRCh38
rs61743313 Chr.2: 21002656-21002656 on Build GRCh38
rs181340304 Chr.2: 21002615-21002615 on Build GRCh38
rs751291501 Chr.2: 21002635-21002635 on Build GRCh38
rs750400586 Chr.2: 21002739-21002739 on Build GRCh38
rs765373860 Chr.2: 21002738-21002738 on Build GRCh38
rs61743299 Chr.2: 21002725-21002725 on Build GRCh38
rs910938141 Chr.2: 21002622-21002622 on Build GRCh38
rs752747528 Chr.2: 21002731-21002731 on Build GRCh38
rs1045669799 Chr.2: 21002676-21002676 on Build GRCh38
rs757398908 Chr.2: 21002652-21002652 on Build GRCh38
rs369527542 Chr.2: 21002734-21002734 on Build GRCh38
rs781061676 Chr.2: 21002643-21002643 on Build GRCh38
rs1801702 Chr.2: 21002613-21002613 on Build GRCh38
rs373945806 Chr.2: 21002612-21002612 on Build GRCh38
rs772581019 Chr.2: 21002718-21002718 on Build GRCh38
rs72654422 Chr.2: 21002619-21002619 on Build GRCh38
rs907126709 Chr.2: 21002683-21002683 on Build GRCh38
rs769409367 Chr.2: 21002707-21002707 on Build GRCh38
rs755429390 Chr.2: 21002766-21002766 on Build GRCh38
rs568054660 Chr.2: 21002743-21002743 on Build GRCh38
rs761276405 Chr.2: 21002616-21002616 on Build GRCh38
rs56675344 Chr.2: 21002723-21002723 on Build GRCh38
rs1052737840 Chr.2: 21002654-21002654 on Build GRCh38
rs776311715 Chr.2: 21002618-21002618 on Build GRCh38
rs777243467 Chr.2: 21002688-21002688 on Build GRCh38
rs749351043 Chr.2: 21002611-21002611 on Build GRCh38
rs767269509 Chr.2: 21002727-21002727 on Build GRCh38
rs748569334 Chr.2: 21002698-21002698 on Build GRCh38
rs780569169 Chr.2: 21002664-21002664 on Build GRCh38
rs372040568 Chr.2: 21002700-21002700 on Build GRCh38
rs139116589 Chr.2: 21002722-21002722 on Build GRCh38
rs145807061 Chr.2: 21002598-21002598 on Build GRCh38
rs746160756 Chr.2: 21002657-21002657 on Build GRCh38
rs540192015 Chr.2: 21002629-21002629 on Build GRCh38
rs12713457 Chr.2: 21002638-21002638 on Build GRCh38
rs377329586 Chr.2: 21002744-21002744 on Build GRCh38
rs556369378 Chr.2: 21002576-21002576 on Build GRCh38
rs563130890 Chr.2: 21002699-21002699 on Build GRCh38
rs200260789 Chr.2: 21002686-21002686 on Build GRCh38
rs61743502 Chr.2: 21002628-21002628 on Build GRCh38
rs766216606 Chr.2: 21002632-21002632 on Build GRCh38
rs758637006 Chr.2: 21002661-21002661 on Build GRCh38
rs758259925 Chr.2: 21002575-21002575 on Build GRCh38
rs919234513 Chr.2: 21002547-21002547 on Build GRCh38
rs769836306 Chr.2: 21002624-21002625 on Build GRCh38
rs764627015 Chr.2: 21002621-21002621 on Build GRCh38
rs371177562 Chr.2: 21002745-21002745 on Build GRCh38
rs143612660 Chr.2: 21002671-21002671 on Build GRCh38
rs1052000878 Chr.2: 21002691-21002691 on Build GRCh38

View Full Product Details