Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52911551-52911810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC2
Entrez Gene ID
55268
Gene Name
enoyl-CoA hydratase domain containing 2
Gene Aliases
-
UniGene
Hs.476319
Gene Chromosome Location
Chr.1: 52895910-52927212 on Build GRCh38
Transcripts
NM_001198961.1 NM_001198962.1 NM_001319958.1 NM_018281.3 XM_011541709.2 XM_011541710.2 XM_011541711.2 XM_011541712.2 XM_011541713.2 XM_011541715.2 XM_011541719.2 XM_011541720.2 XM_011541721.2 XM_011541722.2 XM_011541723.2 XM_011541724.2 XM_011541725.2 XM_011541726.2 XM_011541727.2 XM_017001638.1 XM_017001639.1 XM_017001640.1 XM_017001641.1 XM_017001642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4577437 c.81G>A p.E27E Chr.1: 52911624-52911624 on Build GRCh38
4680078 c.215G>A p.R72Q Chr.1: 52911628-52911628 on Build GRCh38
6651344 c.151_153delTCT p.S51delS Chr.1: 52911759-52911761 on Build GRCh38
4680074 c.102G>T p.L34L Chr.1: 52911603-52911603 on Build GRCh38
4341004 c.126C>T p.G42G Chr.1: 52911579-52911579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758538584 Chr.1: 52911592-52911592 on Build GRCh38
rs1033544304 Chr.1: 52911708-52911708 on Build GRCh38
rs777683028 Chr.1: 52911600-52911600 on Build GRCh38
rs199849538 Chr.1: 52911620-52911620 on Build GRCh38
rs368580572 Chr.1: 52911629-52911629 on Build GRCh38
rs369702852 Chr.1: 52911728-52911728 on Build GRCh38
rs747278747 Chr.1: 52911607-52911607 on Build GRCh38
rs774769180 Chr.1: 52911652-52911653 on Build GRCh38
rs781602712 Chr.1: 52911611-52911611 on Build GRCh38
rs763325358 Chr.1: 52911623-52911623 on Build GRCh38
rs764902748 Chr.1: 52911636-52911636 on Build GRCh38
rs764216740 Chr.1: 52911660-52911660 on Build GRCh38
rs943658773 Chr.1: 52911611-52911612 on Build GRCh38
rs752530332 Chr.1: 52911588-52911588 on Build GRCh38
rs766136963 Chr.1: 52911631-52911631 on Build GRCh38
rs746334615 Chr.1: 52911655-52911656 on Build GRCh38
rs369835429 Chr.1: 52911684-52911684 on Build GRCh38
rs780263189 Chr.1: 52911696-52911696 on Build GRCh38
rs936138222 Chr.1: 52911618-52911618 on Build GRCh38
rs775132834 Chr.1: 52911739-52911739 on Build GRCh38
rs759397766 Chr.1: 52911634-52911634 on Build GRCh38
rs961004778 Chr.1: 52911791-52911791 on Build GRCh38
rs139969883 Chr.1: 52911619-52911619 on Build GRCh38
rs771664696 Chr.1: 52911628-52911628 on Build GRCh38
rs1000700313 Chr.1: 52911707-52911707 on Build GRCh38
rs768183962 Chr.1: 52911681-52911691 on Build GRCh38
rs574010101 Chr.1: 52911650-52911650 on Build GRCh38
rs756950270 Chr.1: 52911579-52911579 on Build GRCh38
rs376597474 Chr.1: 52911672-52911672 on Build GRCh38
rs758245122 Chr.1: 52911659-52911659 on Build GRCh38
rs60877501 Chr.1: 52911676-52911676 on Build GRCh38
rs566778374 Chr.1: 52911711-52911711 on Build GRCh38
rs780846335 Chr.1: 52911632-52911632 on Build GRCh38
rs756529109 Chr.1: 52911677-52911677 on Build GRCh38
rs757347862 Chr.1: 52911755-52911755 on Build GRCh38
rs372614589 Chr.1: 52911661-52911661 on Build GRCh38
rs375548787 Chr.1: 52911652-52911652 on Build GRCh38
rs201328818 Chr.1: 52911706-52911706 on Build GRCh38
rs926105927 Chr.1: 52911578-52911578 on Build GRCh38
rs769043619 Chr.1: 52911702-52911702 on Build GRCh38
rs761464881 Chr.1: 52911771-52911771 on Build GRCh38
rs768996501 Chr.1: 52911754-52911754 on Build GRCh38
rs771128240 Chr.1: 52911610-52911610 on Build GRCh38
rs746232656 Chr.1: 52911616-52911616 on Build GRCh38
rs763014786 Chr.1: 52911741-52911741 on Build GRCh38
rs749682687 Chr.1: 52911585-52911589 on Build GRCh38
rs776049953 Chr.1: 52911702-52911703 on Build GRCh38
rs771294907 Chr.1: 52911640-52911641 on Build GRCh38
rs1017277468 Chr.1: 52911730-52911730 on Build GRCh38
rs140085217 Chr.1: 52911731-52911731 on Build GRCh38
rs757257130 Chr.1: 52911664-52911664 on Build GRCh38
rs376092523 Chr.1: 52911710-52911710 on Build GRCh38
rs749866571 Chr.1: 52911697-52911697 on Build GRCh38
rs764022320 Chr.1: 52911744-52911744 on Build GRCh38

View Full Product Details