Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24117990-24118263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF11
Entrez Gene ID
80344
Gene Name
DDB1 and CUL4 associated factor 11
Gene Aliases
GL014, PRO2389, WDR23
UniGene
Hs.525251
Gene Chromosome Location
Chr.14: 24114697-24125242 on Build GRCh38
Transcripts
NM_001163484.1 NM_025230.4 NM_181357.2 NR_028099.1 NR_028100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477498 c.517C>T p.Q173* Chr.14: 24118095-24118095 on Build GRCh38
1730995 c.477-1G>A p.? Chr.14: 24118054-24118054 on Build GRCh38
5422033 c.508A>G p.S170G Chr.14: 24118086-24118086 on Build GRCh38
3690040 c.533G>A p.G178D Chr.14: 24118111-24118111 on Build GRCh38
5635750 c.537C>A p.I179I Chr.14: 24118115-24118115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201961254 Chr.14: 24118190-24118190 on Build GRCh38
rs759327535 Chr.14: 24118140-24118140 on Build GRCh38
rs759093739 Chr.14: 24118014-24118014 on Build GRCh38
rs948674949 Chr.14: 24118235-24118235 on Build GRCh38
rs963647265 Chr.14: 24118137-24118137 on Build GRCh38
rs899507071 Chr.14: 24118166-24118166 on Build GRCh38
rs191768847 Chr.14: 24118160-24118160 on Build GRCh38
rs1019527939 Chr.14: 24118112-24118112 on Build GRCh38
rs370828167 Chr.14: 24118017-24118017 on Build GRCh38
rs183061328 Chr.14: 24118208-24118208 on Build GRCh38
rs780169242 Chr.14: 24118058-24118058 on Build GRCh38
rs747073593 Chr.14: 24118065-24118065 on Build GRCh38
rs377458609 Chr.14: 24118053-24118053 on Build GRCh38
rs559136856 Chr.14: 24118066-24118066 on Build GRCh38
rs767176066 Chr.14: 24118015-24118015 on Build GRCh38
rs758561532 Chr.14: 24118206-24118206 on Build GRCh38
rs770898555 Chr.14: 24118086-24118086 on Build GRCh38
rs142387074 Chr.14: 24118079-24118079 on Build GRCh38
rs780260343 Chr.14: 24118207-24118207 on Build GRCh38
rs371242058 Chr.14: 24118071-24118071 on Build GRCh38
rs912116879 Chr.14: 24118116-24118116 on Build GRCh38
rs538753994 Chr.14: 24118041-24118041 on Build GRCh38
rs749063211 Chr.14: 24118080-24118080 on Build GRCh38
rs1057495012 Chr.14: 24118213-24118213 on Build GRCh38
rs183350518 Chr.14: 24118029-24118029 on Build GRCh38
rs186195111 Chr.14: 24118150-24118150 on Build GRCh38
rs894739945 Chr.14: 24118228-24118228 on Build GRCh38
rs750688032 Chr.14: 24118034-24118034 on Build GRCh38
rs975061446 Chr.14: 24118155-24118155 on Build GRCh38
rs938944497 Chr.14: 24118210-24118210 on Build GRCh38
rs149132073 Chr.14: 24118236-24118236 on Build GRCh38
rs575893059 Chr.14: 24118119-24118119 on Build GRCh38
rs904591758 Chr.14: 24118237-24118237 on Build GRCh38
rs754320005 Chr.14: 24118169-24118169 on Build GRCh38
rs772041063 Chr.14: 24118111-24118111 on Build GRCh38
rs145287754 Chr.14: 24118189-24118189 on Build GRCh38
rs368549286 Chr.14: 24118171-24118171 on Build GRCh38
rs373152781 Chr.14: 24118019-24118019 on Build GRCh38
rs764618430 Chr.14: 24118164-24118164 on Build GRCh38
rs931087213 Chr.14: 24118183-24118183 on Build GRCh38
rs377657161 Chr.14: 24118023-24118023 on Build GRCh38
rs1000679395 Chr.14: 24118027-24118027 on Build GRCh38
rs576815916 Chr.14: 24118212-24118212 on Build GRCh38
rs754161976 Chr.14: 24118025-24118025 on Build GRCh38
rs774115158 Chr.14: 24118089-24118089 on Build GRCh38

View Full Product Details