Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32949056-32949321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DMA
Entrez Gene ID
3108
Gene Name
major histocompatibility complex, class II, DM alpha
Gene Aliases
D6S222E, DMA, HLADM, RING6
UniGene
Hs.728759
Gene Chromosome Location
Chr.6: 32948614-32953122 on Build GRCh38
Transcripts
NM_006120.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4885980 c.864C>A p.C288* Chr.6: 32949278-32949278 on Build GRCh38
4885979 c.774C>A p.C258* Chr.6: 32949278-32949278 on Build GRCh38
4834879 c.776C>G p.S259* Chr.6: 32949276-32949276 on Build GRCh38
3921497 c.825C>T p.I275I Chr.6: 32949317-32949317 on Build GRCh38
7255257 c.765delG p.K256fs*>6 Chr.6: 32949287-32949287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573793298 Chr.6: 32949144-32949144 on Build GRCh38
rs749995639 Chr.6: 32949285-32949285 on Build GRCh38
rs114253801 Chr.6: 32949265-32949265 on Build GRCh38
rs779711764 Chr.6: 32949289-32949289 on Build GRCh38
rs768293819 Chr.6: 32949226-32949226 on Build GRCh38
rs752124181 Chr.6: 32949268-32949268 on Build GRCh38
rs774366254 Chr.6: 32949234-32949234 on Build GRCh38
rs555544485 Chr.6: 32949083-32949083 on Build GRCh38
rs985100786 Chr.6: 32949128-32949128 on Build GRCh38
rs1046289636 Chr.6: 32949190-32949190 on Build GRCh38
rs919556520 Chr.6: 32949093-32949093 on Build GRCh38
rs376398370 Chr.6: 32949288-32949288 on Build GRCh38
rs759671009 Chr.6: 32949247-32949247 on Build GRCh38
rs908103666 Chr.6: 32949159-32949159 on Build GRCh38
rs776913731 Chr.6: 32949243-32949243 on Build GRCh38
rs781662572 Chr.6: 32949275-32949275 on Build GRCh38
rs577695349 Chr.6: 32949214-32949214 on Build GRCh38
rs765603507 Chr.6: 32949250-32949250 on Build GRCh38
rs781659788 Chr.6: 32949213-32949213 on Build GRCh38
rs771047224 Chr.6: 32949236-32949236 on Build GRCh38
rs748841447 Chr.6: 32949295-32949295 on Build GRCh38
rs373304159 Chr.6: 32949170-32949170 on Build GRCh38
rs757640092 Chr.6: 32949271-32949271 on Build GRCh38
rs763410327 Chr.6: 32949251-32949251 on Build GRCh38
rs550160110 Chr.6: 32949176-32949176 on Build GRCh38
rs1037931907 Chr.6: 32949152-32949152 on Build GRCh38
rs376418557 Chr.6: 32949221-32949221 on Build GRCh38
rs749158231 Chr.6: 32949222-32949222 on Build GRCh38
rs537941911 Chr.6: 32949156-32949156 on Build GRCh38
rs930959719 Chr.6: 32949122-32949122 on Build GRCh38

View Full Product Details