Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157057791-157058052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
UniGene
Hs.129711
Gene Chromosome Location
Chr.5: 156979443-157059119 on Build GRCh38
Transcripts
NM_001173393.2 NM_001308156.1 NM_012206.3 XM_017009339.1 XM_017009340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1672078 c.13G>A p.V5M Chr.5: 157057931-157057931 on Build GRCh38
6546864 c.40C>A p.L14M Chr.5: 157057904-157057904 on Build GRCh38
6546863 c.40C>A p.L14M Chr.5: 157057904-157057904 on Build GRCh38
5646427 c.26G>A p.S9N Chr.5: 157057918-157057918 on Build GRCh38
5646426 c.26G>A p.S9N Chr.5: 157057918-157057918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775401352 Chr.5: 157057882-157057882 on Build GRCh38
rs780795032 Chr.5: 157057910-157057910 on Build GRCh38
rs376702877 Chr.5: 157057874-157057874 on Build GRCh38
rs373333056 Chr.5: 157057992-157057992 on Build GRCh38
rs888073780 Chr.5: 157058013-157058013 on Build GRCh38
rs200455416 Chr.5: 157058008-157058008 on Build GRCh38
rs746142294 Chr.5: 157057950-157057950 on Build GRCh38
rs191484308 Chr.5: 157057947-157057947 on Build GRCh38
rs775186702 Chr.5: 157057923-157057924 on Build GRCh38
rs377610762 Chr.5: 157057949-157057949 on Build GRCh38
rs895692190 Chr.5: 157057972-157057972 on Build GRCh38
rs905509999 Chr.5: 157058003-157058003 on Build GRCh38
rs756741071 Chr.5: 157057925-157057925 on Build GRCh38
rs749655522 Chr.5: 157057861-157057861 on Build GRCh38
rs186293337 Chr.5: 157057897-157057897 on Build GRCh38
rs201144082 Chr.5: 157057867-157057867 on Build GRCh38
rs182096233 Chr.5: 157057852-157057852 on Build GRCh38
rs545225119 Chr.5: 157057853-157057853 on Build GRCh38
rs773143694 Chr.5: 157057990-157057990 on Build GRCh38
rs749757393 Chr.5: 157057937-157057937 on Build GRCh38
rs778418874 Chr.5: 157057932-157057932 on Build GRCh38
rs765149994 Chr.5: 157057892-157057892 on Build GRCh38
rs768658640 Chr.5: 157057989-157057989 on Build GRCh38
rs202130787 Chr.5: 157057896-157057896 on Build GRCh38
rs747588373 Chr.5: 157057914-157057914 on Build GRCh38
rs774730567 Chr.5: 157057869-157057869 on Build GRCh38
rs112079259 Chr.5: 157057981-157057981 on Build GRCh38
rs369161917 Chr.5: 157057893-157057893 on Build GRCh38
rs761864086 Chr.5: 157057889-157057889 on Build GRCh38
rs751243088 Chr.5: 157057909-157057909 on Build GRCh38
rs775701033 Chr.5: 157057978-157057978 on Build GRCh38
rs767667350 Chr.5: 157057875-157057875 on Build GRCh38

View Full Product Details