Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 627224-627451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUL1
Entrez Gene ID
27098
Gene Name
clusterin like 1
Gene Aliases
RA337M
UniGene
Hs.732341
Gene Chromosome Location
Chr.18: 596998-650298 on Build GRCh38
Transcripts
NM_001289036.1 NM_001318522.1 NM_014410.4 NM_199167.1 XM_005258103.3 XM_006722322.2 XM_011525648.2 XM_011525649.1 XM_011525651.2 XM_011525652.1 XM_011525653.2 XM_017025707.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3422284 c.603C>A p.V201V Chr.18: 627276-627276 on Build GRCh38
709110 c.582C>G p.L194L Chr.18: 627255-627255 on Build GRCh38
564627 c.578C>T p.S193F Chr.18: 627251-627251 on Build GRCh38
7361621 c.620A>C p.Q207P Chr.18: 627293-627293 on Build GRCh38
6083414 c.642A>T p.Q214H Chr.18: 627315-627315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775423748 Chr.18: 627343-627343 on Build GRCh38
rs775023261 Chr.18: 627373-627373 on Build GRCh38
rs753466551 Chr.18: 627251-627251 on Build GRCh38
rs754701144 Chr.18: 627263-627263 on Build GRCh38
rs936889229 Chr.18: 627406-627406 on Build GRCh38
rs767965355 Chr.18: 627383-627383 on Build GRCh38
rs548196095 Chr.18: 627381-627381 on Build GRCh38
rs564780184 Chr.18: 627293-627293 on Build GRCh38
rs917274055 Chr.18: 627380-627380 on Build GRCh38
rs765007782 Chr.18: 627396-627396 on Build GRCh38
rs374889105 Chr.18: 627395-627395 on Build GRCh38
rs752090670 Chr.18: 627397-627397 on Build GRCh38
rs532434091 Chr.18: 627295-627295 on Build GRCh38
rs200242838 Chr.18: 627274-627274 on Build GRCh38
rs757232437 Chr.18: 627412-627412 on Build GRCh38
rs764631476 Chr.18: 627266-627266 on Build GRCh38
rs550555452 Chr.18: 627313-627313 on Build GRCh38
rs758005346 Chr.18: 627294-627295 on Build GRCh38
rs115407065 Chr.18: 627315-627315 on Build GRCh38
rs747869709 Chr.18: 627298-627298 on Build GRCh38
rs757925386 Chr.18: 627399-627399 on Build GRCh38
rs1020410544 Chr.18: 627244-627244 on Build GRCh38
rs746053172 Chr.18: 627326-627326 on Build GRCh38
rs199868270 Chr.18: 627388-627388 on Build GRCh38
rs992776128 Chr.18: 627371-627371 on Build GRCh38
rs770109169 Chr.18: 627336-627336 on Build GRCh38
rs566113688 Chr.18: 627408-627408 on Build GRCh38
rs777700996 Chr.18: 627286-627286 on Build GRCh38
rs371155784 Chr.18: 627305-627305 on Build GRCh38
rs780833495 Chr.18: 627420-627420 on Build GRCh38
rs763977404 Chr.18: 627403-627403 on Build GRCh38
rs769232173 Chr.18: 627369-627369 on Build GRCh38
rs766077524 Chr.18: 627250-627250 on Build GRCh38
rs986404246 Chr.18: 627382-627382 on Build GRCh38
rs752310151 Chr.18: 627273-627273 on Build GRCh38
rs529629931 Chr.18: 627325-627325 on Build GRCh38
rs920944448 Chr.18: 627355-627355 on Build GRCh38

View Full Product Details