Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35756768-35757017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB6
Entrez Gene ID
126393
Gene Name
heat shock protein family B (small) member 6
Gene Aliases
HEL55, Hsp20, PPP1R91
UniGene
Hs.534538
Gene Chromosome Location
Chr.19: 35754566-35757029 on Build GRCh38
Transcripts
NM_144617.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5696512 c.79C>T p.R27C Chr.19: 35756930-35756930 on Build GRCh38
7371581 c.59C>T p.P20L Chr.19: 35756950-35756950 on Build GRCh38
5474137 c.27G>A p.P9P Chr.19: 35756982-35756982 on Build GRCh38
5562842 c.16C>T p.P6S Chr.19: 35756993-35756993 on Build GRCh38
6524890 c.68C>T p.S23L Chr.19: 35756941-35756941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11549029 Chr.19: 35756950-35756950 on Build GRCh38
rs758088660 Chr.19: 35756795-35756795 on Build GRCh38
rs1004843934 Chr.19: 35756896-35756896 on Build GRCh38
rs960170560 Chr.19: 35756900-35756901 on Build GRCh38
rs62109459 Chr.19: 35756909-35756909 on Build GRCh38
rs34531131 Chr.19: 35756799-35756800 on Build GRCh38
rs376421880 Chr.19: 35756955-35756955 on Build GRCh38
rs1005618052 Chr.19: 35756832-35756832 on Build GRCh38
rs62109458 Chr.19: 35756904-35756904 on Build GRCh38
rs1040901928 Chr.19: 35756954-35756954 on Build GRCh38
rs542529846 Chr.19: 35756990-35756990 on Build GRCh38
rs200536955 Chr.19: 35756980-35756980 on Build GRCh38
rs912589768 Chr.19: 35756952-35756952 on Build GRCh38
rs758796871 Chr.19: 35756959-35756959 on Build GRCh38
rs996023208 Chr.19: 35756910-35756910 on Build GRCh38
rs756278720 Chr.19: 35756869-35756869 on Build GRCh38
rs182616636 Chr.19: 35756934-35756934 on Build GRCh38
rs888174982 Chr.19: 35756984-35756984 on Build GRCh38
rs970296172 Chr.19: 35756854-35756854 on Build GRCh38
rs750670441 Chr.19: 35756802-35756802 on Build GRCh38
rs1002502974 Chr.19: 35756793-35756793 on Build GRCh38
rs777357750 Chr.19: 35756930-35756944 on Build GRCh38
rs911253870 Chr.19: 35756948-35756948 on Build GRCh38
rs1016087084 Chr.19: 35756901-35756901 on Build GRCh38
rs979166419 Chr.19: 35756941-35756941 on Build GRCh38
rs954275211 Chr.19: 35756945-35756945 on Build GRCh38
rs755529886 Chr.19: 35756939-35756939 on Build GRCh38
rs766478369 Chr.19: 35756874-35756874 on Build GRCh38
rs913293563 Chr.19: 35756915-35756915 on Build GRCh38
rs538390060 Chr.19: 35756889-35756889 on Build GRCh38
rs1005340898 Chr.19: 35756998-35756998 on Build GRCh38
rs886347933 Chr.19: 35756881-35756881 on Build GRCh38
rs989049311 Chr.19: 35756964-35756964 on Build GRCh38
rs1023329634 Chr.19: 35756846-35756846 on Build GRCh38

View Full Product Details