Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27147557-27147782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA-AS3
Entrez Gene ID
100133311
Gene Name
HOXA cluster antisense RNA 3
Gene Aliases
HOXA6as
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27140364-27155928 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2 NM_024014.3

Gene Symbol
HOXA3
Entrez Gene ID
3200
Gene Name
homeobox A3
Gene Aliases
HOX1, HOX1E
UniGene
Hs.659337
Gene Chromosome Location
Chr.7: 27106190-27152581 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2 NM_024014.3

Gene Symbol
HOXA6
Entrez Gene ID
3203
Gene Name
homeobox A6
Gene Aliases
HOX1, HOX1.2, HOX1B
UniGene
Hs.679517
Gene Chromosome Location
Chr.7: 27145583-27147774 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 XM_005249730.2 XM_006715715.2 XM_011515343.2 NM_024014.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746310 c.97G>A p.D33N Chr.7: 27147653-27147653 on Build GRCh38
81276 c.18G>A p.V6V Chr.7: 27147732-27147732 on Build GRCh38
5680921 c.68G>T p.G23V Chr.7: 27147682-27147682 on Build GRCh38
3027258 c.42T>G p.L14L Chr.7: 27147708-27147708 on Build GRCh38
3027259 c.33C>T p.P11P Chr.7: 27147717-27147717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779478368 Chr.7: 27147587-27147587 on Build GRCh38
rs935855248 Chr.7: 27147756-27147756 on Build GRCh38
rs767355550 Chr.7: 27147637-27147637 on Build GRCh38
rs767727688 Chr.7: 27147717-27147717 on Build GRCh38
rs758058929 Chr.7: 27147753-27147753 on Build GRCh38
rs763114537 Chr.7: 27147622-27147622 on Build GRCh38
rs774078821 Chr.7: 27147599-27147599 on Build GRCh38
rs753858296 Chr.7: 27147728-27147728 on Build GRCh38
rs760180498 Chr.7: 27147606-27147606 on Build GRCh38
rs747316620 Chr.7: 27147602-27147602 on Build GRCh38
rs754568432 Chr.7: 27147655-27147655 on Build GRCh38
rs372988125 Chr.7: 27147748-27147748 on Build GRCh38
rs376057404 Chr.7: 27147649-27147649 on Build GRCh38
rs750229263 Chr.7: 27147646-27147646 on Build GRCh38
rs748838301 Chr.7: 27147589-27147589 on Build GRCh38
rs759402904 Chr.7: 27147710-27147710 on Build GRCh38
rs760426520 Chr.7: 27147579-27147580 on Build GRCh38
rs764297599 Chr.7: 27147691-27147691 on Build GRCh38
rs764200063 Chr.7: 27147623-27147623 on Build GRCh38
rs770373490 Chr.7: 27147686-27147686 on Build GRCh38
rs373980441 Chr.7: 27147624-27147624 on Build GRCh38
rs911772148 Chr.7: 27147635-27147635 on Build GRCh38
rs765854235 Chr.7: 27147616-27147616 on Build GRCh38
rs944658409 Chr.7: 27147645-27147645 on Build GRCh38
rs777590094 Chr.7: 27147754-27147754 on Build GRCh38
rs370341498 Chr.7: 27147702-27147702 on Build GRCh38
rs371909980 Chr.7: 27147719-27147719 on Build GRCh38
rs866015529 Chr.7: 27147672-27147672 on Build GRCh38
rs778399764 Chr.7: 27147658-27147658 on Build GRCh38
rs762909337 Chr.7: 27147688-27147688 on Build GRCh38
rs766149827 Chr.7: 27147619-27147620 on Build GRCh38
rs370785785 Chr.7: 27147720-27147720 on Build GRCh38
rs951345958 Chr.7: 27147605-27147605 on Build GRCh38
rs752270376 Chr.7: 27147752-27147752 on Build GRCh38
rs771463001 Chr.7: 27147603-27147603 on Build GRCh38
rs754442674 Chr.7: 27147738-27147738 on Build GRCh38
rs776564519 Chr.7: 27147685-27147687 on Build GRCh38
rs757038196 Chr.7: 27147625-27147625 on Build GRCh38
rs149022222 Chr.7: 27147681-27147681 on Build GRCh38
rs878913509 Chr.7: 27147590-27147590 on Build GRCh38
rs890054197 Chr.7: 27147700-27147700 on Build GRCh38
rs771409970 Chr.7: 27147678-27147678 on Build GRCh38
rs375214907 Chr.7: 27147725-27147725 on Build GRCh38
rs768156585 Chr.7: 27147598-27147598 on Build GRCh38
rs922795801 Chr.7: 27147633-27147633 on Build GRCh38
rs776113892 Chr.7: 27147687-27147687 on Build GRCh38
rs747901674 Chr.7: 27147674-27147674 on Build GRCh38
rs186744061 Chr.7: 27147647-27147647 on Build GRCh38
rs780074710 Chr.7: 27147648-27147648 on Build GRCh38
rs746211230 Chr.7: 27147760-27147760 on Build GRCh38

View Full Product Details