Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64542278-64542529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA2
Entrez Gene ID
3306
Gene Name
heat shock protein family A (Hsp70) member 2
Gene Aliases
HSP70-2, HSP70-3
UniGene
Hs.432648
Gene Chromosome Location
Chr.14: 64540468-64543237 on Build GRCh38
Transcripts
NM_021979.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1300759 c.1656C>T p.I552I Chr.14: 64542505-64542505 on Build GRCh38
6682585 c.1540A>C p.S514R Chr.14: 64542389-64542389 on Build GRCh38
7422936 c.1576G>A p.E526K Chr.14: 64542425-64542425 on Build GRCh38
1300758 c.1444G>A p.D482N Chr.14: 64542293-64542293 on Build GRCh38
7280711 c.1470C>T p.N490N Chr.14: 64542319-64542319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375156060 Chr.14: 64542461-64542461 on Build GRCh38
rs774308215 Chr.14: 64542383-64542383 on Build GRCh38
rs139711462 Chr.14: 64542448-64542448 on Build GRCh38
rs763879452 Chr.14: 64542470-64542470 on Build GRCh38
rs762927698 Chr.14: 64542358-64542358 on Build GRCh38
rs985886722 Chr.14: 64542492-64542492 on Build GRCh38
rs190645540 Chr.14: 64542373-64542373 on Build GRCh38
rs11551976 Chr.14: 64542502-64542502 on Build GRCh38
rs559245797 Chr.14: 64542503-64542503 on Build GRCh38
rs769297658 Chr.14: 64542326-64542326 on Build GRCh38
rs202207348 Chr.14: 64542333-64542333 on Build GRCh38
rs142119907 Chr.14: 64542349-64542349 on Build GRCh38
rs369473748 Chr.14: 64542410-64542410 on Build GRCh38
rs759421405 Chr.14: 64542386-64542386 on Build GRCh38
rs764742296 Chr.14: 64542427-64542427 on Build GRCh38
rs781645807 Chr.14: 64542331-64542331 on Build GRCh38
rs745990927 Chr.14: 64542457-64542457 on Build GRCh38
rs200575249 Chr.14: 64542428-64542428 on Build GRCh38
rs770107690 Chr.14: 64542347-64542347 on Build GRCh38
rs776299096 Chr.14: 64542479-64542479 on Build GRCh38
rs1011001299 Chr.14: 64542445-64542445 on Build GRCh38
rs201636500 Chr.14: 64542391-64542391 on Build GRCh38
rs770898950 Chr.14: 64542448-64542449 on Build GRCh38
rs146374519 Chr.14: 64542417-64542417 on Build GRCh38
rs372954597 Chr.14: 64542437-64542437 on Build GRCh38
rs536762997 Chr.14: 64542409-64542409 on Build GRCh38
rs781774342 Chr.14: 64542435-64542435 on Build GRCh38
rs756861833 Chr.14: 64542421-64542421 on Build GRCh38
rs780416765 Chr.14: 64542301-64542301 on Build GRCh38
rs747478968 Chr.14: 64542313-64542313 on Build GRCh38
rs45545942 Chr.14: 64542508-64542508 on Build GRCh38
rs45447398 Chr.14: 64542335-64542335 on Build GRCh38
rs761476742 Chr.14: 64542481-64542481 on Build GRCh38
rs556559367 Chr.14: 64542438-64542438 on Build GRCh38
rs17853551 Chr.14: 64542439-64542439 on Build GRCh38
rs576414293 Chr.14: 64542456-64542456 on Build GRCh38
rs375115167 Chr.14: 64542395-64542395 on Build GRCh38
rs760461731 Chr.14: 64542469-64542469 on Build GRCh38
rs867605397 Chr.14: 64542401-64542401 on Build GRCh38
rs749469619 Chr.14: 64542446-64542446 on Build GRCh38
rs758029974 Chr.14: 64542504-64542504 on Build GRCh38
rs775467719 Chr.14: 64542462-64542462 on Build GRCh38

View Full Product Details