Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3659899-3660170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART1
Entrez Gene ID
417
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ART2, ARTC1, CD296, RT6
UniGene
Hs.382188
Gene Chromosome Location
Chr.11: 3645110-3665595 on Build GRCh38
Transcripts
NM_004314.2 XM_011520114.2 XM_017017763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6345247 c.489G>T p.Q163H Chr.11: 3660008-3660008 on Build GRCh38
4748938 c.389A>G p.E130G Chr.11: 3659908-3659908 on Build GRCh38
6537183 c.545A>G p.H182R Chr.11: 3660064-3660064 on Build GRCh38
4856397 c.535C>T p.R179* Chr.11: 3660054-3660054 on Build GRCh38
4921221 c.634C>T p.Q212* Chr.11: 3660153-3660153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767771808 Chr.11: 3659982-3659982 on Build GRCh38
rs202112440 Chr.11: 3660122-3660122 on Build GRCh38
rs991136254 Chr.11: 3659972-3659972 on Build GRCh38
rs1044310817 Chr.11: 3660079-3660094 on Build GRCh38
rs111942933 Chr.11: 3659939-3659939 on Build GRCh38
rs762197868 Chr.11: 3660037-3660037 on Build GRCh38
rs141732093 Chr.11: 3660028-3660028 on Build GRCh38
rs750686385 Chr.11: 3659987-3659987 on Build GRCh38
rs148703944 Chr.11: 3659926-3659926 on Build GRCh38
rs774844448 Chr.11: 3660085-3660085 on Build GRCh38
rs781422161 Chr.11: 3659922-3659922 on Build GRCh38
rs888495221 Chr.11: 3659919-3659919 on Build GRCh38
rs765406319 Chr.11: 3660102-3660102 on Build GRCh38
rs371263931 Chr.11: 3660066-3660066 on Build GRCh38
rs577558078 Chr.11: 3659968-3659968 on Build GRCh38
rs761164939 Chr.11: 3660145-3660145 on Build GRCh38
rs141183632 Chr.11: 3659991-3659991 on Build GRCh38
rs202219245 Chr.11: 3660087-3660087 on Build GRCh38
rs542871399 Chr.11: 3660073-3660073 on Build GRCh38
rs554173168 Chr.11: 3659995-3659995 on Build GRCh38
rs771324525 Chr.11: 3660150-3660150 on Build GRCh38
rs145152353 Chr.11: 3660015-3660015 on Build GRCh38
rs772525381 Chr.11: 3660089-3660089 on Build GRCh38
rs757425295 Chr.11: 3659921-3659921 on Build GRCh38
rs761598555 Chr.11: 3659933-3659933 on Build GRCh38
rs750734969 Chr.11: 3660043-3660043 on Build GRCh38
rs757189893 Chr.11: 3660121-3660121 on Build GRCh38
rs145405723 Chr.11: 3660054-3660054 on Build GRCh38
rs748021891 Chr.11: 3660010-3660010 on Build GRCh38
rs773884895 Chr.11: 3660138-3660138 on Build GRCh38
rs927702624 Chr.11: 3660091-3660091 on Build GRCh38
rs142006583 Chr.11: 3660093-3660093 on Build GRCh38
rs746683411 Chr.11: 3660021-3660021 on Build GRCh38
rs769388795 Chr.11: 3660082-3660082 on Build GRCh38
rs754871942 Chr.11: 3660005-3660005 on Build GRCh38
rs537517512 Chr.11: 3659937-3659937 on Build GRCh38
rs777796511 Chr.11: 3660117-3660117 on Build GRCh38
rs760968477 Chr.11: 3660094-3660094 on Build GRCh38
rs143216187 Chr.11: 3660069-3660069 on Build GRCh38
rs766036791 Chr.11: 3659944-3659944 on Build GRCh38
rs147920431 Chr.11: 3660020-3660020 on Build GRCh38
rs201487637 Chr.11: 3660000-3660000 on Build GRCh38
rs745586244 Chr.11: 3660124-3660124 on Build GRCh38
rs752681438 Chr.11: 3660110-3660110 on Build GRCh38
rs138825846 Chr.11: 3660079-3660079 on Build GRCh38
rs754155931 Chr.11: 3660097-3660097 on Build GRCh38
rs151146245 Chr.11: 3659984-3659984 on Build GRCh38
rs892156505 Chr.11: 3660115-3660115 on Build GRCh38
rs749144608 Chr.11: 3660055-3660055 on Build GRCh38
rs779643913 Chr.11: 3660131-3660131 on Build GRCh38
rs545730767 Chr.11: 3659974-3659974 on Build GRCh38
rs776376314 Chr.11: 3660116-3660117 on Build GRCh38
rs760397227 Chr.11: 3659943-3659943 on Build GRCh38
rs748155690 Chr.11: 3660068-3660068 on Build GRCh38
rs759029297 Chr.11: 3660029-3660029 on Build GRCh38
rs774042346 Chr.11: 3659932-3659932 on Build GRCh38
rs759245792 Chr.11: 3660119-3660120 on Build GRCh38
rs199699575 Chr.11: 3660023-3660023 on Build GRCh38
rs374138196 Chr.11: 3660072-3660072 on Build GRCh38
rs138349653 Chr.11: 3660148-3660148 on Build GRCh38
rs368030263 Chr.11: 3660130-3660130 on Build GRCh38
rs376054375 Chr.11: 3659963-3659963 on Build GRCh38
rs770721141 Chr.11: 3660076-3660076 on Build GRCh38
rs769221472 Chr.11: 3660006-3660006 on Build GRCh38
rs748677669 Chr.11: 3660132-3660132 on Build GRCh38
rs757702310 Chr.11: 3659975-3659975 on Build GRCh38
rs759645257 Chr.11: 3660101-3660101 on Build GRCh38
rs753925166 Chr.11: 3660056-3660056 on Build GRCh38
rs772551570 Chr.11: 3660030-3660030 on Build GRCh38
rs1010554048 Chr.11: 3660127-3660127 on Build GRCh38
rs145683174 Chr.11: 3660137-3660137 on Build GRCh38
rs200211996 Chr.11: 3659949-3659949 on Build GRCh38
rs767921923 Chr.11: 3660038-3660038 on Build GRCh38
rs113817011 Chr.11: 3660090-3660090 on Build GRCh38
rs149229160 Chr.11: 3660065-3660065 on Build GRCh38
rs543916143 Chr.11: 3660088-3660088 on Build GRCh38
rs751530347 Chr.11: 3660120-3660120 on Build GRCh38
rs774993469 Chr.11: 3660036-3660036 on Build GRCh38
rs561441207 Chr.11: 3660078-3660078 on Build GRCh38
rs150462911 Chr.11: 3660033-3660033 on Build GRCh38
rs371356884 Chr.11: 3659925-3659925 on Build GRCh38
rs758416985 Chr.11: 3660111-3660111 on Build GRCh38

View Full Product Details