Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49718359-49718616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO3
Entrez Gene ID
386724
Gene Name
adhesion molecule with Ig like domain 3
Gene Aliases
ALI3, AMIGO-3
UniGene
Hs.567903
Gene Chromosome Location
Chr.3: 49716834-49719805 on Build GRCh38
Transcripts
NM_198722.2 NM_022064.4 NR_135218.1 XM_006713285.1 XM_011533995.1 XM_017007018.1

Gene Symbol
RNF123
Entrez Gene ID
63891
Gene Name
ring finger protein 123
Gene Aliases
FP1477, KPC1
UniGene
Hs.553723
Gene Chromosome Location
Chr.3: 49688753-49721529 on Build GRCh38
Transcripts
NM_198722.2 NM_022064.4 NR_135218.1 XM_006713285.1 XM_011533995.1 XM_017007018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599258 c.851C>T p.P284L Chr.3: 49718615-49718615 on Build GRCh38
6599260 c.898T>C p.C300R Chr.3: 49718568-49718568 on Build GRCh38
4518923 c.877_878GG>AA p.G293N Chr.3: 49718588-49718589 on Build GRCh38
6599259 c.1039G>A p.A347T Chr.3: 49718427-49718427 on Build GRCh38
1182678 c.948G>T p.Q316H Chr.3: 49718518-49718518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018486205 Chr.3: 49718492-49718492 on Build GRCh38
rs772464782 Chr.3: 49718393-49718393 on Build GRCh38
rs867957408 Chr.3: 49718468-49718468 on Build GRCh38
rs748525963 Chr.3: 49718594-49718594 on Build GRCh38
rs777353932 Chr.3: 49718466-49718466 on Build GRCh38
rs776880189 Chr.3: 49718412-49718412 on Build GRCh38
rs373689365 Chr.3: 49718598-49718598 on Build GRCh38
rs780494082 Chr.3: 49718477-49718477 on Build GRCh38
rs762999839 Chr.3: 49718520-49718520 on Build GRCh38
rs573198411 Chr.3: 49718449-49718449 on Build GRCh38
rs1030585913 Chr.3: 49718524-49718524 on Build GRCh38
rs1051795677 Chr.3: 49718415-49718415 on Build GRCh38
rs779537959 Chr.3: 49718382-49718382 on Build GRCh38
rs376736320 Chr.3: 49718441-49718441 on Build GRCh38
rs752548201 Chr.3: 49718540-49718540 on Build GRCh38
rs761004181 Chr.3: 49718398-49718398 on Build GRCh38
rs755772643 Chr.3: 49718552-49718552 on Build GRCh38
rs774191576 Chr.3: 49718531-49718531 on Build GRCh38
rs1041997599 Chr.3: 49718395-49718395 on Build GRCh38
rs368808320 Chr.3: 49718471-49718471 on Build GRCh38
rs903154052 Chr.3: 49718404-49718404 on Build GRCh38
rs755865197 Chr.3: 49718450-49718450 on Build GRCh38
rs540880083 Chr.3: 49718580-49718580 on Build GRCh38
rs989014753 Chr.3: 49718588-49718588 on Build GRCh38
rs747369013 Chr.3: 49718485-49718485 on Build GRCh38
rs766332423 Chr.3: 49718529-49718529 on Build GRCh38
rs889194560 Chr.3: 49718423-49718423 on Build GRCh38
rs868789247 Chr.3: 49718544-49718544 on Build GRCh38
rs748825526 Chr.3: 49718469-49718469 on Build GRCh38
rs765250301 Chr.3: 49718425-49718425 on Build GRCh38
rs199566932 Chr.3: 49718394-49718394 on Build GRCh38
rs768918710 Chr.3: 49718407-49718407 on Build GRCh38
rs981834081 Chr.3: 49718498-49718498 on Build GRCh38
rs778101475 Chr.3: 49718448-49718449 on Build GRCh38
rs780864622 Chr.3: 49718442-49718442 on Build GRCh38
rs62262720 Chr.3: 49718392-49718392 on Build GRCh38
rs753397985 Chr.3: 49718562-49718562 on Build GRCh38
rs780590724 Chr.3: 49718579-49718579 on Build GRCh38
rs34810303 Chr.3: 49718533-49718533 on Build GRCh38
rs150551790 Chr.3: 49718589-49718589 on Build GRCh38
rs768952020 Chr.3: 49718503-49718503 on Build GRCh38
rs781467596 Chr.3: 49718590-49718590 on Build GRCh38
rs756757861 Chr.3: 49718576-49718576 on Build GRCh38
rs761982260 Chr.3: 49718505-49718505 on Build GRCh38
rs766239635 Chr.3: 49718437-49718437 on Build GRCh38
rs138809316 Chr.3: 49718515-49718515 on Build GRCh38
rs750531309 Chr.3: 49718434-49718434 on Build GRCh38
rs962582792 Chr.3: 49718495-49718495 on Build GRCh38
rs749799985 Chr.3: 49718473-49718486 on Build GRCh38
rs762882013 Chr.3: 49718436-49718436 on Build GRCh38
rs767368457 Chr.3: 49718538-49718538 on Build GRCh38
rs539432112 Chr.3: 49718388-49718388 on Build GRCh38
rs761861468 Chr.3: 49718419-49718419 on Build GRCh38
rs199670077 Chr.3: 49718440-49718440 on Build GRCh38

View Full Product Details