Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134932897-134933138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALD1
Entrez Gene ID
800
Gene Name
caldesmon 1
Gene Aliases
CDM, H-CAD, HCAD, L-CAD, LCAD, NAG22
UniGene
Hs.490203
Gene Chromosome Location
Chr.7: 134779413-134970729 on Build GRCh38
Transcripts
NM_004342.6 NM_033138.3 NM_033139.3 NM_033140.3 NM_033157.3 XM_017012650.1 XM_017012651.1 XM_017012652.1 XM_017012653.1 XM_017012654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134577 c.299G>C p.R100T Chr.7: 134933086-134933086 on Build GRCh38
7352552 c.364G>A p.D122N Chr.7: 134933133-134933133 on Build GRCh38
6432847 c.296G>A p.R99H Chr.7: 134933065-134933065 on Build GRCh38
7137270 c.336G>A p.Q112Q Chr.7: 134933123-134933123 on Build GRCh38
4185702 c.295C>A p.R99S Chr.7: 134933064-134933064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34385239 Chr.7: 134933101-134933101 on Build GRCh38
rs1035398218 Chr.7: 134932992-134932992 on Build GRCh38
rs780105261 Chr.7: 134932936-134932936 on Build GRCh38
rs190269973 Chr.7: 134933119-134933119 on Build GRCh38
rs931252688 Chr.7: 134932924-134932924 on Build GRCh38
rs918406497 Chr.7: 134932921-134932921 on Build GRCh38
rs761604756 Chr.7: 134933028-134933028 on Build GRCh38
rs753306157 Chr.7: 134933059-134933059 on Build GRCh38
rs748916759 Chr.7: 134932965-134932965 on Build GRCh38
rs143639360 Chr.7: 134933065-134933065 on Build GRCh38
rs984073728 Chr.7: 134932944-134932944 on Build GRCh38
rs372544119 Chr.7: 134932970-134932970 on Build GRCh38
rs148090500 Chr.7: 134933107-134933107 on Build GRCh38
rs757031720 Chr.7: 134932938-134932938 on Build GRCh38
rs771601905 Chr.7: 134933074-134933074 on Build GRCh38
rs781128570 Chr.7: 134932948-134932948 on Build GRCh38
rs763146732 Chr.7: 134933090-134933090 on Build GRCh38
rs772919992 Chr.7: 134933098-134933098 on Build GRCh38
rs772334697 Chr.7: 134932979-134932979 on Build GRCh38
rs550549442 Chr.7: 134932968-134932968 on Build GRCh38
rs748464902 Chr.7: 134932974-134932974 on Build GRCh38
rs767285471 Chr.7: 134933037-134933037 on Build GRCh38
rs372385136 Chr.7: 134933097-134933097 on Build GRCh38
rs143238463 Chr.7: 134932958-134932958 on Build GRCh38
rs759796889 Chr.7: 134933013-134933013 on Build GRCh38
rs542527726 Chr.7: 134933043-134933043 on Build GRCh38
rs766257245 Chr.7: 134933118-134933118 on Build GRCh38
rs759636510 Chr.7: 134933077-134933077 on Build GRCh38
rs780938122 Chr.7: 134932940-134932940 on Build GRCh38
rs755648979 Chr.7: 134932952-134932952 on Build GRCh38
rs148922576 Chr.7: 134933050-134933050 on Build GRCh38
rs147912516 Chr.7: 134932998-134932998 on Build GRCh38
rs765513603 Chr.7: 134933017-134933017 on Build GRCh38
rs772479657 Chr.7: 134933070-134933070 on Build GRCh38
rs375816775 Chr.7: 134933073-134933073 on Build GRCh38
rs1046037633 Chr.7: 134933024-134933024 on Build GRCh38
rs375796865 Chr.7: 134932997-134932997 on Build GRCh38
rs770000559 Chr.7: 134933083-134933083 on Build GRCh38
rs752987758 Chr.7: 134933025-134933025 on Build GRCh38
rs778481089 Chr.7: 134933064-134933064 on Build GRCh38
rs778056399 Chr.7: 134933072-134933072 on Build GRCh38
rs779508995 Chr.7: 134933051-134933051 on Build GRCh38
rs775853439 Chr.7: 134933088-134933088 on Build GRCh38
rs776895371 Chr.7: 134933002-134933002 on Build GRCh38
rs369898278 Chr.7: 134933076-134933076 on Build GRCh38
rs750335210 Chr.7: 134932951-134932951 on Build GRCh38
rs749896298 Chr.7: 134932966-134932968 on Build GRCh38

View Full Product Details