Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95081383-95081634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD6
Entrez Gene ID
55785
Gene Name
FYVE, RhoGEF and PH domain containing 6
Gene Aliases
ZFYVE24
UniGene
Hs.506381
Gene Chromosome Location
Chr.12: 95076749-95217464 on Build GRCh38
Transcripts
NM_018351.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777129 c.4264G>C p.E1422Q Chr.12: 95081549-95081549 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371185019 Chr.12: 95081455-95081455 on Build GRCh38
rs900097314 Chr.12: 95081479-95081479 on Build GRCh38
rs374356534 Chr.12: 95081435-95081436 on Build GRCh38
rs770416049 Chr.12: 95081480-95081480 on Build GRCh38
rs368337004 Chr.12: 95081470-95081470 on Build GRCh38
rs761950535 Chr.12: 95081571-95081571 on Build GRCh38
rs775802779 Chr.12: 95081528-95081528 on Build GRCh38
rs774404322 Chr.12: 95081483-95081483 on Build GRCh38
rs757415670 Chr.12: 95081453-95081453 on Build GRCh38
rs372013977 Chr.12: 95081554-95081554 on Build GRCh38
rs746332961 Chr.12: 95081504-95081504 on Build GRCh38
rs557408624 Chr.12: 95081464-95081464 on Build GRCh38
rs764210145 Chr.12: 95081560-95081560 on Build GRCh38
rs186209784 Chr.12: 95081605-95081605 on Build GRCh38
rs773737870 Chr.12: 95081487-95081487 on Build GRCh38
rs777544940 Chr.12: 95081467-95081467 on Build GRCh38
rs979142901 Chr.12: 95081433-95081433 on Build GRCh38
rs765324635 Chr.12: 95081574-95081574 on Build GRCh38
rs925482372 Chr.12: 95081443-95081443 on Build GRCh38
rs74485804 Chr.12: 95081588-95081588 on Build GRCh38
rs752533756 Chr.12: 95081604-95081604 on Build GRCh38
rs776854986 Chr.12: 95081565-95081565 on Build GRCh38
rs765059339 Chr.12: 95081458-95081458 on Build GRCh38
rs772588408 Chr.12: 95081512-95081512 on Build GRCh38
rs781088469 Chr.12: 95081463-95081463 on Build GRCh38
rs796737246 Chr.12: 95081536-95081536 on Build GRCh38
rs1048921816 Chr.12: 95081586-95081586 on Build GRCh38
rs755000020 Chr.12: 95081462-95081462 on Build GRCh38
rs1007778110 Chr.12: 95081603-95081603 on Build GRCh38
rs77727695 Chr.12: 95081444-95081444 on Build GRCh38
rs750672123 Chr.12: 95081582-95081582 on Build GRCh38
rs79415743 Chr.12: 95081575-95081575 on Build GRCh38

View Full Product Details