Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114835987-114836248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B1
Entrez Gene ID
57700
Gene Name
family with sequence similarity 160 member B1
Gene Aliases
KIAA1600, bA106M7.3
UniGene
Hs.192619
Gene Chromosome Location
Chr.10: 114821744-114899827 on Build GRCh38
Transcripts
NM_001135051.1 NM_020940.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011840 c.425T>C p.V142A Chr.10: 114836149-114836149 on Build GRCh38
683232 c.502C>A p.L168M Chr.10: 114836226-114836226 on Build GRCh38
4011841 c.425T>C p.V142A Chr.10: 114836149-114836149 on Build GRCh38
6518309 c.476G>A p.C159Y Chr.10: 114836200-114836200 on Build GRCh38
683230 c.519A>G p.L173L Chr.10: 114836243-114836243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546636573 Chr.10: 114836063-114836063 on Build GRCh38
rs760018864 Chr.10: 114836138-114836138 on Build GRCh38
rs748025753 Chr.10: 114836075-114836075 on Build GRCh38
rs765690848 Chr.10: 114836141-114836141 on Build GRCh38
rs766081469 Chr.10: 114836119-114836119 on Build GRCh38
rs528762320 Chr.10: 114836040-114836040 on Build GRCh38
rs776400804 Chr.10: 114836132-114836132 on Build GRCh38
rs771627903 Chr.10: 114836087-114836087 on Build GRCh38
rs764236262 Chr.10: 114836151-114836151 on Build GRCh38
rs781437291 Chr.10: 114836176-114836176 on Build GRCh38
rs753303555 Chr.10: 114836148-114836148 on Build GRCh38
rs751550246 Chr.10: 114836153-114836153 on Build GRCh38
rs543087902 Chr.10: 114836025-114836025 on Build GRCh38
rs201143653 Chr.10: 114836082-114836082 on Build GRCh38
rs778582871 Chr.10: 114836181-114836181 on Build GRCh38
rs368403731 Chr.10: 114836109-114836109 on Build GRCh38
rs754524961 Chr.10: 114836157-114836157 on Build GRCh38
rs927483371 Chr.10: 114836221-114836221 on Build GRCh38
rs866430088 Chr.10: 114836161-114836161 on Build GRCh38
rs1019592098 Chr.10: 114836143-114836143 on Build GRCh38
rs750643308 Chr.10: 114836203-114836203 on Build GRCh38
rs375064220 Chr.10: 114836099-114836099 on Build GRCh38
rs976923013 Chr.10: 114836069-114836069 on Build GRCh38
rs754828533 Chr.10: 114836204-114836204 on Build GRCh38
rs1027037174 Chr.10: 114836215-114836215 on Build GRCh38
rs1052899467 Chr.10: 114836053-114836053 on Build GRCh38
rs890348005 Chr.10: 114836101-114836101 on Build GRCh38
rs909543308 Chr.10: 114836111-114836111 on Build GRCh38
rs1025376519 Chr.10: 114836076-114836076 on Build GRCh38
rs951030853 Chr.10: 114836077-114836077 on Build GRCh38
rs192320674 Chr.10: 114836217-114836217 on Build GRCh38

View Full Product Details