Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49859542-49859805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKV
Entrez Gene ID
79012
Gene Name
CaM kinase like vesicle associated
Gene Aliases
1G5, VACAMKL
UniGene
Hs.145156
Gene Chromosome Location
Chr.3: 49857981-49870222 on Build GRCh38
Transcripts
NM_001320147.1 NM_024046.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4648717 c.1158C>A p.A386A Chr.3: 49859666-49859666 on Build GRCh38
4668973 c.1184C>T p.T395I Chr.3: 49859640-49859640 on Build GRCh38
6619336 c.1150C>T p.R384C Chr.3: 49859674-49859674 on Build GRCh38
4625130 c.1046G>A p.G349E Chr.3: 49859778-49859778 on Build GRCh38
6619342 c.1030G>A p.D344N Chr.3: 49859794-49859794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200719095 Chr.3: 49859578-49859578 on Build GRCh38
rs753686285 Chr.3: 49859591-49859591 on Build GRCh38
rs778259877 Chr.3: 49859641-49859641 on Build GRCh38
rs774277628 Chr.3: 49859569-49859569 on Build GRCh38
rs764079591 Chr.3: 49859685-49859685 on Build GRCh38
rs759724043 Chr.3: 49859573-49859573 on Build GRCh38
rs748648023 Chr.3: 49859637-49859637 on Build GRCh38
rs757869703 Chr.3: 49859625-49859625 on Build GRCh38
rs868127460 Chr.3: 49859683-49859683 on Build GRCh38
rs779164652 Chr.3: 49859787-49859787 on Build GRCh38
rs756675433 Chr.3: 49859761-49859761 on Build GRCh38
rs756724824 Chr.3: 49859639-49859639 on Build GRCh38
rs779453635 Chr.3: 49859630-49859630 on Build GRCh38
rs757819063 Chr.3: 49859607-49859630 on Build GRCh38
rs763848470 Chr.3: 49859589-49859589 on Build GRCh38
rs756893325 Chr.3: 49859605-49859605 on Build GRCh38
rs866327852 Chr.3: 49859643-49859643 on Build GRCh38
rs767496171 Chr.3: 49859576-49859576 on Build GRCh38
rs749812937 Chr.3: 49859779-49859779 on Build GRCh38
rs774562098 Chr.3: 49859665-49859665 on Build GRCh38
rs150720881 Chr.3: 49859680-49859680 on Build GRCh38
rs749727173 Chr.3: 49859655-49859655 on Build GRCh38
rs201137978 Chr.3: 49859574-49859574 on Build GRCh38
rs751164217 Chr.3: 49859747-49859747 on Build GRCh38
rs913640257 Chr.3: 49859725-49859748 on Build GRCh38
rs765879384 Chr.3: 49859743-49859743 on Build GRCh38
rs750044422 Chr.3: 49859717-49859717 on Build GRCh38
rs772138978 Chr.3: 49859679-49859679 on Build GRCh38
rs934957635 Chr.3: 49859651-49859651 on Build GRCh38
rs758016459 Chr.3: 49859740-49859740 on Build GRCh38
rs764938176 Chr.3: 49859606-49859606 on Build GRCh38
rs947599904 Chr.3: 49859681-49859681 on Build GRCh38
rs1053448441 Chr.3: 49859674-49859674 on Build GRCh38
rs746089971 Chr.3: 49859673-49859673 on Build GRCh38
rs760588166 Chr.3: 49859684-49859684 on Build GRCh38
rs149916894 Chr.3: 49859730-49859730 on Build GRCh38
rs1020690637 Chr.3: 49859699-49859699 on Build GRCh38
rs142041712 Chr.3: 49859662-49859662 on Build GRCh38
rs377281206 Chr.3: 49859710-49859710 on Build GRCh38
rs760641215 Chr.3: 49859587-49859587 on Build GRCh38
rs556114266 Chr.3: 49859623-49859623 on Build GRCh38
rs778486705 Chr.3: 49859765-49859765 on Build GRCh38
rs371059995 Chr.3: 49859780-49859780 on Build GRCh38
rs199584164 Chr.3: 49859709-49859709 on Build GRCh38

View Full Product Details