Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 410226-410471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR2
Entrez Gene ID
26063
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Gene Aliases
PDCR, SDR17C1
UniGene
Hs.628831
Gene Chromosome Location
Chr.16: 401858-412487 on Build GRCh38
Transcripts
NM_020664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7218695 c.453G>T p.K151N Chr.16: 410358-410358 on Build GRCh38
3361766 c.634T>G p.*212G Chr.16: 410352-410352 on Build GRCh38
7213479 c.338-5T>C p.? Chr.16: 410238-410238 on Build GRCh38
1377882 c.551G>A p.R184H Chr.16: 410269-410269 on Build GRCh38
6642128 c.461G>A p.R154Q Chr.16: 410366-410366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772183628 Chr.16: 410261-410261 on Build GRCh38
rs374863109 Chr.16: 410353-410353 on Build GRCh38
rs771190349 Chr.16: 410352-410352 on Build GRCh38
rs144593130 Chr.16: 410250-410250 on Build GRCh38
rs756055283 Chr.16: 410341-410341 on Build GRCh38
rs559915457 Chr.16: 410252-410252 on Build GRCh38
rs150670530 Chr.16: 410365-410365 on Build GRCh38
rs770300567 Chr.16: 410319-410319 on Build GRCh38
rs3743891 Chr.16: 410415-410415 on Build GRCh38
rs771806477 Chr.16: 410419-410419 on Build GRCh38
rs188648786 Chr.16: 410425-410425 on Build GRCh38
rs749261930 Chr.16: 410440-410440 on Build GRCh38
rs376506167 Chr.16: 410345-410345 on Build GRCh38
rs377244160 Chr.16: 410386-410386 on Build GRCh38
rs374254424 Chr.16: 410251-410251 on Build GRCh38
rs996639929 Chr.16: 410340-410340 on Build GRCh38
rs747251416 Chr.16: 410350-410350 on Build GRCh38
rs758046162 Chr.16: 410352-410388 on Build GRCh38
rs201481606 Chr.16: 410333-410333 on Build GRCh38
rs778034146 Chr.16: 410346-410346 on Build GRCh38
rs774605806 Chr.16: 410392-410420 on Build GRCh38
rs3743892 Chr.16: 410448-410448 on Build GRCh38
rs752994223 Chr.16: 410351-410351 on Build GRCh38
rs768491642 Chr.16: 410365-410368 on Build GRCh38
rs772287975 Chr.16: 410310-410310 on Build GRCh38
rs201553914 Chr.16: 410383-410383 on Build GRCh38
rs200583305 Chr.16: 410403-410403 on Build GRCh38
rs942707966 Chr.16: 410331-410331 on Build GRCh38
rs761302440 Chr.16: 410370-410370 on Build GRCh38
rs763774191 Chr.16: 410323-410323 on Build GRCh38
rs1027133091 Chr.16: 410344-410344 on Build GRCh38
rs371859454 Chr.16: 410338-410338 on Build GRCh38
rs74944003 Chr.16: 410269-410269 on Build GRCh38
rs542989271 Chr.16: 410299-410299 on Build GRCh38
rs774052111 Chr.16: 410368-410368 on Build GRCh38
rs199661265 Chr.16: 410382-410382 on Build GRCh38
rs116164566 Chr.16: 410274-410274 on Build GRCh38
rs554529207 Chr.16: 410276-410276 on Build GRCh38
rs748445439 Chr.16: 410255-410255 on Build GRCh38
rs922870459 Chr.16: 410435-410435 on Build GRCh38
rs201367889 Chr.16: 410407-410407 on Build GRCh38
rs769768839 Chr.16: 410287-410287 on Build GRCh38
rs769206831 Chr.16: 410416-410416 on Build GRCh38
rs1047348456 Chr.16: 410445-410445 on Build GRCh38
rs751042971 Chr.16: 410290-410290 on Build GRCh38
rs934803979 Chr.16: 410306-410306 on Build GRCh38
rs757988805 Chr.16: 410289-410289 on Build GRCh38
rs759970750 Chr.16: 410316-410316 on Build GRCh38
rs759622204 Chr.16: 410412-410413 on Build GRCh38
rs745338566 Chr.16: 410350-410351 on Build GRCh38
rs1005267803 Chr.16: 410364-410364 on Build GRCh38
rs368586364 Chr.16: 410247-410247 on Build GRCh38
rs748095831 Chr.16: 410382-410384 on Build GRCh38
rs531767849 Chr.16: 410388-410388 on Build GRCh38
rs779472210 Chr.16: 410420-410420 on Build GRCh38
rs760691987 Chr.16: 410268-410268 on Build GRCh38
rs760291085 Chr.16: 410380-410380 on Build GRCh38
rs773281208 Chr.16: 410263-410263 on Build GRCh38
rs558262846 Chr.16: 410301-410301 on Build GRCh38
rs780160364 Chr.16: 410342-410342 on Build GRCh38
rs565878000 Chr.16: 410417-410417 on Build GRCh38
rs768964049 Chr.16: 410366-410366 on Build GRCh38
rs763342513 Chr.16: 410322-410322 on Build GRCh38
rs79060096 Chr.16: 410358-410358 on Build GRCh38
rs780529988 Chr.16: 410408-410408 on Build GRCh38
rs141151070 Chr.16: 410246-410246 on Build GRCh38
rs765659217 Chr.16: 410277-410277 on Build GRCh38
rs575650329 Chr.16: 410292-410292 on Build GRCh38
rs769804320 Chr.16: 410383-410390 on Build GRCh38
rs908677978 Chr.16: 410446-410446 on Build GRCh38
rs982606099 Chr.16: 410354-410354 on Build GRCh38
rs746834253 Chr.16: 410360-410395 on Build GRCh38
rs1007853040 Chr.16: 410317-410317 on Build GRCh38
rs1039250947 Chr.16: 410427-410427 on Build GRCh38
rs767288280 Chr.16: 410378-410378 on Build GRCh38
rs371311313 Chr.16: 410379-410379 on Build GRCh38
rs201491114 Chr.16: 410335-410335 on Build GRCh38
rs183925447 Chr.16: 410314-410314 on Build GRCh38
rs779621533 Chr.16: 410358-410358 on Build GRCh38
rs368053751 Chr.16: 410355-410355 on Build GRCh38
rs527313474 Chr.16: 410376-410376 on Build GRCh38
rs776471619 Chr.16: 410370-410402 on Build GRCh38
rs369453909 Chr.16: 410402-410402 on Build GRCh38
rs550183591 Chr.16: 410397-410397 on Build GRCh38
rs773335348 Chr.16: 410313-410313 on Build GRCh38
rs778376604 Chr.16: 410274-410288 on Build GRCh38
rs532650826 Chr.16: 410412-410412 on Build GRCh38
rs751223310 Chr.16: 410327-410327 on Build GRCh38
rs548488938 Chr.16: 410452-410452 on Build GRCh38
rs576518923 Chr.16: 410302-410302 on Build GRCh38
rs752549486 Chr.16: 410395-410395 on Build GRCh38
rs759870779 Chr.16: 410275-410275 on Build GRCh38
rs77446963 Chr.16: 410321-410321 on Build GRCh38
rs536028257 Chr.16: 410426-410426 on Build GRCh38

View Full Product Details