Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110070387-110070622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX3
Entrez Gene ID
257
Gene Name
ALX homeobox 3
Gene Aliases
FND, FND1
UniGene
Hs.669953
Gene Chromosome Location
Chr.1: 110060375-110070700 on Build GRCh38
Transcripts
NM_006492.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328004 c.130C>T p.R44C Chr.1: 110070483-110070483 on Build GRCh38
4004678 c.79G>A p.G27S Chr.1: 110070534-110070534 on Build GRCh38
4632542 c.181C>A p.L61I Chr.1: 110070432-110070432 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1019628352 Chr.1: 110070568-110070568 on Build GRCh38
rs771353213 Chr.1: 110070434-110070434 on Build GRCh38
rs774644842 Chr.1: 110070476-110070476 on Build GRCh38
rs777537752 Chr.1: 110070419-110070420 on Build GRCh38
rs372506798 Chr.1: 110070422-110070422 on Build GRCh38
rs893323237 Chr.1: 110070521-110070521 on Build GRCh38
rs775462465 Chr.1: 110070453-110070453 on Build GRCh38
rs754614088 Chr.1: 110070420-110070420 on Build GRCh38
rs781525090 Chr.1: 110070438-110070438 on Build GRCh38
rs375790592 Chr.1: 110070458-110070458 on Build GRCh38
rs1020856559 Chr.1: 110070530-110070530 on Build GRCh38
rs967688577 Chr.1: 110070532-110070532 on Build GRCh38
rs1001916056 Chr.1: 110070465-110070465 on Build GRCh38
rs545925004 Chr.1: 110070409-110070409 on Build GRCh38
rs757496737 Chr.1: 110070447-110070447 on Build GRCh38
rs769009378 Chr.1: 110070470-110070470 on Build GRCh38
rs975514659 Chr.1: 110070594-110070594 on Build GRCh38
rs1011770350 Chr.1: 110070529-110070529 on Build GRCh38
rs964012645 Chr.1: 110070575-110070575 on Build GRCh38
rs555884259 Chr.1: 110070433-110070433 on Build GRCh38
rs12756321 Chr.1: 110070411-110070411 on Build GRCh38
rs931112658 Chr.1: 110070419-110070419 on Build GRCh38
rs746339603 Chr.1: 110070550-110070550 on Build GRCh38

View Full Product Details