Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145730933-145730708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD160
Entrez Gene ID
11126
Gene Name
CD160 molecule
Gene Aliases
BY55, NK1, NK28
UniGene
Hs.488237
Gene Chromosome Location
Chr.1: 145719312-145739288 on Build GRCh38
Transcripts
NM_007053.3 NR_103845.1 XM_005272929.3 XM_011509104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873450 c.218G>A p.S73N Chr.1: 145730888-145730888 on Build GRCh38
5377514 c.242G>A p.R81K Chr.1: 145730912-145730912 on Build GRCh38
5553721 c.211G>C p.E71Q Chr.1: 145730881-145730881 on Build GRCh38
1626388 c.251G>A p.G84E Chr.1: 145730921-145730921 on Build GRCh38
5377515 c.164G>A p.G55E Chr.1: 145730834-145730834 on Build GRCh38
View More

View Full Product Details