Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35973302-35973567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGO3
Entrez Gene ID
192669
Gene Name
argonaute 3, RISC catalytic component
Gene Aliases
EIF2C3
UniGene
Hs.657659
Gene Chromosome Location
Chr.1: 35925198-36072037 on Build GRCh38
Transcripts
NM_024852.3 NM_177422.2 XM_005270575.3 XM_011540879.2 XM_017000523.1 XM_017000528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5173261 c.542_543insA p.F182fs*9 Chr.1: 35973395-35973396 on Build GRCh38
4921449 c.631A>G p.M211V Chr.1: 35973484-35973484 on Build GRCh38
4925111 c.589A>T p.R197W Chr.1: 35973442-35973442 on Build GRCh38
4769648 c.654C>T p.I218I Chr.1: 35973507-35973507 on Build GRCh38
4877096 c.623G>A p.R208Q Chr.1: 35973476-35973476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189379458 Chr.1: 35973532-35973532 on Build GRCh38
rs771632440 Chr.1: 35973405-35973405 on Build GRCh38
rs777956533 Chr.1: 35973392-35973392 on Build GRCh38
rs755135833 Chr.1: 35973334-35973334 on Build GRCh38
rs910634817 Chr.1: 35973340-35973340 on Build GRCh38
rs966645450 Chr.1: 35973359-35973359 on Build GRCh38
rs756414637 Chr.1: 35973536-35973536 on Build GRCh38
rs982934585 Chr.1: 35973382-35973382 on Build GRCh38
rs367796824 Chr.1: 35973424-35973424 on Build GRCh38
rs1057132464 Chr.1: 35973520-35973520 on Build GRCh38
rs772943203 Chr.1: 35973507-35973507 on Build GRCh38
rs775034538 Chr.1: 35973448-35973448 on Build GRCh38
rs147480778 Chr.1: 35973426-35973426 on Build GRCh38
rs752784137 Chr.1: 35973346-35973346 on Build GRCh38
rs762768828 Chr.1: 35973526-35973526 on Build GRCh38
rs781292616 Chr.1: 35973335-35973335 on Build GRCh38
rs143581788 Chr.1: 35973475-35973475 on Build GRCh38
rs761675753 Chr.1: 35973495-35973495 on Build GRCh38
rs768040424 Chr.1: 35973478-35973478 on Build GRCh38
rs776275393 Chr.1: 35973484-35973484 on Build GRCh38
rs753968144 Chr.1: 35973543-35973543 on Build GRCh38
rs765172603 Chr.1: 35973503-35973503 on Build GRCh38
rs756118563 Chr.1: 35973390-35973390 on Build GRCh38
rs779353364 Chr.1: 35973410-35973410 on Build GRCh38
rs532521981 Chr.1: 35973534-35973534 on Build GRCh38
rs749743973 Chr.1: 35973397-35973397 on Build GRCh38
rs764075067 Chr.1: 35973541-35973541 on Build GRCh38

View Full Product Details