Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134357574-134357847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
UniGene
Hs.426312
Gene Chromosome Location
Chr.3: 134355345-134375417 on Build GRCh38
Transcripts
NM_001278683.1 NM_001278685.1 NM_016201.3 XM_006713654.2 XM_011512881.1 XM_017006580.1 XM_017006581.1 XM_017006582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4570071 c.2324T>A p.M775K Chr.3: 134357724-134357724 on Build GRCh38
4922575 c.2341T>G p.*781G Chr.3: 134357707-134357707 on Build GRCh38
5398206 c.2316G>T p.L772F Chr.3: 134357732-134357732 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753843754 Chr.3: 134357681-134357682 on Build GRCh38
rs896993017 Chr.3: 134357663-134357663 on Build GRCh38
rs918589716 Chr.3: 134357613-134357613 on Build GRCh38
rs757748021 Chr.3: 134357610-134357610 on Build GRCh38
rs773545914 Chr.3: 134357761-134357761 on Build GRCh38
rs762200254 Chr.3: 134357748-134357748 on Build GRCh38
rs1011329782 Chr.3: 134357814-134357814 on Build GRCh38
rs778798799 Chr.3: 134357795-134357795 on Build GRCh38
rs765644346 Chr.3: 134357759-134357759 on Build GRCh38
rs138701132 Chr.3: 134357745-134357745 on Build GRCh38
rs1004982873 Chr.3: 134357631-134357631 on Build GRCh38
rs191988589 Chr.3: 134357674-134357674 on Build GRCh38
rs1057154295 Chr.3: 134357626-134357626 on Build GRCh38
rs747721662 Chr.3: 134357811-134357811 on Build GRCh38
rs142696435 Chr.3: 134357664-134357664 on Build GRCh38
rs760230621 Chr.3: 134357659-134357680 on Build GRCh38
rs948250748 Chr.3: 134357667-134357667 on Build GRCh38
rs745688180 Chr.3: 134357798-134357798 on Build GRCh38
rs757110162 Chr.3: 134357716-134357716 on Build GRCh38
rs570666973 Chr.3: 134357617-134357617 on Build GRCh38
rs757147755 Chr.3: 134357685-134357685 on Build GRCh38
rs763732073 Chr.3: 134357786-134357786 on Build GRCh38
rs895877119 Chr.3: 134357627-134357627 on Build GRCh38
rs749069936 Chr.3: 134357703-134357703 on Build GRCh38
rs745831847 Chr.3: 134357731-134357731 on Build GRCh38
rs763549585 Chr.3: 134357767-134357767 on Build GRCh38
rs546488524 Chr.3: 134357666-134357666 on Build GRCh38
rs376825279 Chr.3: 134357739-134357739 on Build GRCh38
rs368864039 Chr.3: 134357778-134357778 on Build GRCh38
rs777056670 Chr.3: 134357747-134357747 on Build GRCh38
rs753496383 Chr.3: 134357791-134357791 on Build GRCh38
rs778583451 Chr.3: 134357757-134357758 on Build GRCh38
rs747133356 Chr.3: 134357741-134357741 on Build GRCh38
rs766839769 Chr.3: 134357769-134357769 on Build GRCh38
rs755736379 Chr.3: 134357691-134357691 on Build GRCh38
rs373799192 Chr.3: 134357788-134357788 on Build GRCh38
rs1052753982 Chr.3: 134357768-134357768 on Build GRCh38
rs937328346 Chr.3: 134357625-134357625 on Build GRCh38
rs750942114 Chr.3: 134357684-134357684 on Build GRCh38
rs781668635 Chr.3: 134357668-134357668 on Build GRCh38
rs746432080 Chr.3: 134357687-134357687 on Build GRCh38
rs763584864 Chr.3: 134357661-134357663 on Build GRCh38
rs780353480 Chr.3: 134357723-134357723 on Build GRCh38
rs550519624 Chr.3: 134357605-134357605 on Build GRCh38
rs931355637 Chr.3: 134357634-134357634 on Build GRCh38
rs752254423 Chr.3: 134357690-134357690 on Build GRCh38
rs772003555 Chr.3: 134357737-134357737 on Build GRCh38
rs770344425 Chr.3: 134357700-134357700 on Build GRCh38
rs927604720 Chr.3: 134357618-134357618 on Build GRCh38
rs913024239 Chr.3: 134357809-134357809 on Build GRCh38
rs754385200 Chr.3: 134357686-134357686 on Build GRCh38
rs778651035 Chr.3: 134357725-134357725 on Build GRCh38
rs752185703 Chr.3: 134357774-134357774 on Build GRCh38

View Full Product Details